Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Dispensing Pumps  
Dispensing Pumps
  More Information Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
Masterflex Peristaltic Pumps
• Complete dispensing systems with Tygon LFL tubing – up and running within minutes!
• Flow rate 28 to 1,700 ml/min
Peristaltic pump system
• Includes remote control options
from  Call for price   More details
Diaphragm Metering Pump
• Rugged, chemically resistant casing with choice of wetted parts
• Select from manual, remote or digital control models
Diaphragm metering pump
• Flow ranges: Liquid 0.4 ml/min – 76 LPH
from  Call for price   More details
Cole-Parmer Syringe Pumps
• Ideal for low-flow, low pressure precision applications
• Flow rates up to 500 ml/hr
syringe pump
• Programmable models available
from  £990.00   More details
Piston Pump Heads
• High accuracy - better than +/- 1%
• Flow rates up to 1035 ml/min available
piston pump
• Order drives seperately
from  £193.00   More details
Modular Digital Dispensing Drive
• Designed for accurate dispensing or general fluid transfer
• Flow rates for 0.01 to 26 LPM
brushless peristaltic pump
• Brushless motor virtually eliminates maintenance
from  £2828.00   More details
Cole-Parmer UK’s dispensing pumps offer a high degree of accuracy and are used in a wide variety of applications where controlled dispensing of liquid is required. For dispensing set volumes from nano litres to litres Cole-Parmer’s range of dispensing pumps can dispense at flow rates from 1 nL/min to 45 LPM with pressure options to 17.3 bar.
Cole-Parmer UK offers a range of dispensing pump technologies, including piston pumps, peristaltic pumps, diaphragm pumps and syringe pumps to allow us to offer you the best solution for your application. Whether your application is laboratory, pilot plant, industrial or OEM based Cole-Parmer has over 50 years of application experience. Browse through our on-line catalogue which includes detailed pump specification or contact our Application Specialists for free technical support.
piston pump
Piston Pumps
Ideal for laboratory, process, industrial and OEM applications these pumps offer high accuracy and excellent repeatability.
  • Flow: 0.15 ml/min to 2.3 LPM
  • Max Pressure: 6.8 bar
  • Typical Dispensing Accuracy: ±1%
  • Best repeatability of Cole-Parmer dispenser pumps
  • Long periods between maintenance make these pumps suitable for continuous duty
  • Pump heads are available for connection to a wide choice of drives and controllers including dispense by volume, RS-232 control
peristaltic pump
Peristaltic Pumps
Widely used in laboratory, process and OEM applications including chemical, food, pharmaceutical, semiconductor and water treatment industries
  • Flow: 0.0006 ml/min to 45 LPM
  • Max Pressure: 8.6 bar
  • Typical Dispensing Accuracy: ±0.5%
  • Program dispensing parameters include dispense volumes, number of cycles dispensed and interval delay times
  • Fluid does not contact any part of the pump except the tubing
diaphragm pump
Diaphragm Pumps
Widely used in industrial applications including dispensing of chemicals in process applications, water and waste water treatment
  • Flow: 0.4 ml/min to 8.3 LPM
  • Max Pressure: 17.3 bar
  • Typical Dispensing Accuracy: ±2%
  • Programmable models feature external control modes – pulse multiply, pulse divide or 4-20mA response to control dispensing.
syringe pump
Syringe Pump
Widely used in laboratory for nanofluidic and microfluidic infusion and withdrawal applications
  • Flow: 1 nL/min to 147 ml/min
  • Typical Dispensing Accuracy: ±0.5%
  • Provides high accuracy delivery of ultra low flows
  • Built-in program allows you to enter up to eight commands – with the option of repeating commands to allow you to program flow rates, hold, pause and change directions
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007