Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Featured Products
Masterflex Peristaltic Pumps
Peristaltic pump system
  • Complete dispensing systems with Tygon LFL tubing - up and running within minutes!
  • Flow rate 28 to 1,700 ml/min
  • Includes remote control options
from $1402.00 More details
Diaphragm Metering Pump
Diaphragm metering pump
  • Rugged, chemically resistant casing with choice of wetted parts
  • Select from manual, remote or digital control models
  • Flow ranges: Liquid 0.4 ml/min - 76 LPH
from $536.00 More details
Single Syringe Infusion Pump
syringe pump
  • Microprocessor motor control and precision gearing
  • Obtain precise flow rates down to 0.2µL/hour to 500ml/hour
  • Accepts syringes from 10µL to 60ml
from $1371.00 More details
Centrifugal Magnetic Drive Pumps
Centrifugal Magnetic Drive Pumps
  • Internal cooling circuit reduces bearing temperature
  • Flow rates from 8 LPM to 134 LPM
  • Enclosed motor for use in harsh environments
from $212.00 More details
Atex-Approved Stainless Steel Double-Diaphragm Pumps
atex aod pumps
  • Flow rates of 400 LPM
  • Viscosities up to 11,000 cp
  • Prevents electro-static discharge that could ignite in a gaseous explosive environment
from $3104.00 More details
Complete Drum and Container Pumps
drum pump
  • System includes tube and motor - tube material options ensure chemical compatability
  • Ability to run dry
  • Flow rates up to 120 LPM
from $805.00 More details
Siphon/Lift Pail Pump
hand operated pump
  • Suitable for pumping petroleum products or water
  • Cut inlet tube to fit your container
  • Maximum suction lift is 10ft
from $34.00 More details
Industrial Gear Pump
industrial gear pump
  • Bidirectional pump to allow easy change of flow direction
  • Able to pump fluids with viscosities of up to 100 centipoise (cp)
  • Flow rate up to 220 LPM
from $1447.00 More details
Cole-Parmer Catalogue > Transfer Pumps
Transfer Pumps
Cole-Parmer offers an unrivalled choice of transfer pumps - whether you are looking for pumping technology, chemical pumps, liquid pumps, water pumps or non viscous fluids at high or low flow rates, Cole-Parmer offers the technology and experience to help.
Cole-Parmer's wide range of transfer pump technologies includes; Air Operated Double Diaphragm Pumps (AODD Pumps), Centrifugal pumps, Diaphragm pumps, Drum pumps, Gear pumps, Hand pumps, Peristaltic pumps and Submersible pumps. Our pumps are able to offer flow rates up to 2200 LPM and pressures up to 25 bar.
Whether you are involved in an experimental and research laboratory, industrial production or original equipment manufacturing (OEM), Cole-Parmer has the right transfer pump for your application. Browse through our on-line catalogue which includes detailed specification or contact our Application Specialists for free technical support.
plastic hand operated drum pump
Hand Pumps
Available in a range of container adapters and styles including water hand pumps, heavy duty acid transfer pumps, FDA compliant hand pumps, stainless steel hand pumps and petrol hand pumps
  • Flow: up to 75 LPM (20 GPM)
  • Suction Lift: up to 3.7m (12')
Search for hand pumps on Cole-Parmer's On-line Catalogue
magnetic drive centrifugal pump
Centrifugal Pumps
Offer high flow rates and low pressures, and pulseless flow at a low price - they are ideal for your general transfer applications. Our range includes low cost centrifugal pumps for water transfer and chemically resistant models for highly corrosive chemicals.
  • Flow: up to 2200 LPM
  • Max Pressure: 25 bar
Search for centrifugal pumps on Cole-Parmer's On-line Catalogue
drum and container pump
Drum Pumps
Cole-Parmer drum pumps offer choice of material and length of the tube set to match your application need including general purpose, high viscosity and sanitary sets. Complete the system by selecting an electric of air operated motor.
  • Flow: up to 240 LPM
  • Max Pressure: 2.1 bar
Search for drum pumps on Cole-Parmer's On-line Catalogue
Submersible Pump with Carrying Handle
Submersible Pumps
Wide choice available including low cost plastic submersible pumps for emptying of pools and stainless steel sewage pumps
  • Flow: up to 500 LPM
  • Max Pressure: 1.6 bar
Search for submersible pumps on Cole-Parmer's On-line Catalogue
air operated double diaphragm pumps
Air Operated Double Diaphragm Pumps (AODD Pumps)
Ideal when transferring media with particulates, these pumps can handle up to 90% solids and have a low shear effect. Pumps can be run dry without damage to them
  • Flow: up to 480 LPM
  • Max Pressure: 8 bar
Search for AODD pumps on Cole-Parmer's On-line Catalogue
diaphragm pumps
Diaphragm Pumps
These low cost utility pumps are ideal for general transfer of fluids. Many models include pressure switches to protect the pump from damage
  • Flow: up to 60 LPM
  • Max Pressure: 30 bar
Search for diaphragm pumps on Cole-Parmer's On-line Catalogue
peristaltic pumps
Peristaltic Pumps
Media remains in the tubing allowing for simple setup and low maintenance
  • Flow: up to 45 LPM
  • Max Pressure: 8.6 bar
Search for peristaltic pumps on Cole-Parmer's On-line Catalogue
image4
Gear Pumps
Ideal for the transfer of polymers and corrosive chemicals gear pumps are well suited for the transfer of viscous liquids
  • Flow: up to 189 LPM
  • Max Pressure: 8.6 bar
Search for gear pumps on Cole-Parmer's On-line Catalogue
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish