Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Featured Products
Masterflex Peristaltic Pumps
Peristaltic pump system
  • Complete dispensing systems with Tygon LFL tubing - up and running within minutes!
  • Flow rate 28 to 1,700 ml/min
  • Includes remote control options
from $1402.00 More details
Diaphragm Metering Pump
Diaphragm metering pump
  • Rugged, chemically resistant casing with choice of wetted parts
  • Select from manual, remote or digital control models
  • Flow ranges: Liquid 0.4 ml/min - 76 LPH
from $536.00 More details
Single Syringe Infusion Pump
syringe pump
  • Microprocessor motor control and precision gearing
  • Obtain precise flow rates down to 0.2µL/hour to 500ml/hour
  • Accepts syringes from 10µL to 60ml
from $1371.00 More details
Centrifugal Magnetic Drive Pumps
Centrifugal Magnetic Drive Pumps
  • Internal cooling circuit reduces bearing temperature
  • Flow rates from 8 LPM to 134 LPM
  • Enclosed motor for use in harsh environments
from $212.00 More details
Valveless Piston Pump Heads
valveless piston pump head
  • Pump fluids with viscosities up to 500 cp
  • Flow range available for 0 to 1296 ml/min
  • Low dead volume
from $480.00 More details
Ismatec High-Accuracy Low Flow Pumps
ismatec pump
  • Up to 24 high accuracy channels in one pump
  • Analogue input and output controls
  • Flow rate per channel - 0.0012 to 8.86ml/min
from $3553.00 More details
Masterflex C/L Varible - Speed Tubing Pumps
CL pumps
  • Flow range: 0.001 to 39.4 ml/min depending on rpm and tubing size
  • Remove control connections on back of pump; start/stop pump with contact closure
  • Built-in tubing retainers firmly grip the tubing - change tubing quickly and easily
from $598.00 More details
Programmable Multi-Syringe Pumps
multichannel pump
  • Program flow rate hold, pause and chnage of directions
  • Ten Syringes Pump
  • Flow rate per syringe 0.001µL/hr to 147ml/min
from $3965.00 More details
Air Admiral Vacuum/Pressure pumps and stations
vacuum pump
  • Compact size and integral handle make pump easily transportable
  • Low-friction piston runs cool and quiet for a long service life with a consistent performance.
  • Free air capacity of 10.5LPM
from $300.00 More details
Corrosion Resistant Vacuum Pumps
vacuum pump2
  • Quiet, reliable pumps with excellent chemical compatibility
  • Free air capacity - 5.5 LPM
  • Max Vacuum 25.2
from $525.00 More details
Rotary Vane Vacuum Pumps
vacuum pump 3
  • Compact design - half the size of a typical rotary vane pump
  • Two stage rotary vane vacuum pumps are designed to reduce the number of wear parts
  • Free air capacity of 47 LPM and vacuum of 2 x 10-3 mbar
from $2335.00 More details
Cole-Parmer Catalogue > Pumps > Laboratory pumps
Laboratory Pumps and Lab Dosing Pumps
Cole-Parmer offers an extensive range of laboratory pumps. Cole-Parmer has lab dosing pumps able to provide highly accurate control for nanofluidic and microfluidic infusion, laboratory pumps to dose media, lab pumps for general transfer of media and a range laboratory diaphragm and rotary vacuum pumps. Offering flow rates from 1 nL/min to hundreds of litres per minute with pressure options up to 345 bar, and vacuum less than 10 -3 mbar.
Browse our on-line catalogue or alternatively contact our Application Specialists for free technical support and let us bring over 50 years of laboratory pumping experience to help solve your application needs.
Cole-Parmer's lab pumps can be split in to three main categories:
Laboratory Metering Pumps - for media applications requiring a high degree of accuracy
Laboratory Transfer Pumps - for media applications requiring general transfer of liquid
Laboratory Vacuum Pumps - for vacuum applications
Laboratory Metering Pumps
Laboratory metering pumps offer a high degree of accuracy and are used in a wide variety of applications where precise volumes of liquid are required to be moved in a specified time. Used regularly in the lab, these lab pumps are commonly used to feed chromatography systems and accurately dose controlled amounts of chemicals in to reactor chambers.
Available in flow rates offering control from 1 nL/min upwards the three most popular types of lab metering pumps are peristaltic pumps, piston pumps and syringe pumps:
image1
Peristaltic Pumps
One of the easiest pumps to use - the system can be setup in minutes and as the media is confined to the tubing offers high purity and easy, non-specialised maintenance.
  • Flow: 0.0006 ml/min to 45 LPM
  • Max Pressure: 8.6 bar
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232
  • Media is confined to the tubing allowing contamination free pumping and easy maintenance, minimising down time.
image2
Piston Pump
Piston pumps offer high accuracy and repeatability over long periods and are ideal for use in high pressure delivery of media including HPLC pumping.
  • Flow: 0.04 ml/min to 1.3 LPM
  • Max Pressure: 345 bar
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232
  • Highly accurate over long periods of operation and provide excellent repeatability
image3
Syringe Pump
Widely used in laboratory for nanofluidic and microfluidic infusion and withdrawal applications
  • Flow: 1 nL/min to 147 ml/min
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232.
  • Provides high accuracy delivery of ultra low flows
See our metering pump section for a full list of metering pump technologies.
Laboratory Transfer Pumps
Laboratory transfer pumps are used for general purpose transfer applications such as cleaning of equipment, transfer of chemicals from drums to smaller bottles. Popular pumping technologies include centrifugal pumps, diaphragm pumps and peristaltic pumps.
Centrifugal Magnetic
Centrifugal Pumps
Low cost solution these pumps are ideal for transfer and recirculation of clean liquids.
  • Flow: 1 LPM to 2,200 LPM
  • Max Pressure: 25 bar
  • Offers pulseless flow and available in a wide range of materials
Moderate Flow Positi
Diaphragm Pump
These types of diaphragm pumps are known as positive displacement pumps as they deliver a definite volume of liquid using one or two flexible diaphragms for each cycle of pump operation. Ideal for general transfer and spraying applications, our range can offer flow rates of up to 64 LPM, pressures up to 30 bar and with material options to allow compatibility with corrosive liquids.
  • Flow rates to 64 LPM
  • Pressures up to 30 bar
  • Use for general transfer and spraying applications
image4
Peristaltic Pumps
One of the easiest pumps to use – the system can be setup in minutes and as the media is confined to the tubing offers high purity and easy, non-specialised maintenance.
  • Flow: 0.0006 ml/min to 45 LPM
  • Max Pressure: 8.6 bar
  • Media is confined to the tubing allowing contamination free pumping and easy maintenance, minimising down time.
See our transfer pump section for a full list of transfer pump technologies.
Laboratory Vacuum Pumps
Cole-Parmer offer a range of vacuum diaphragm pumps for vacuum levels down to 0.3 mbar and rotary vacuum pumps for vacuum levels down to 10-3 mbar.
Vacuubrand <small><sup>®</sup></small>  diap
Laboratory Diaphragm Vacuum Pumps
Diaphragm pumps offer low ultimate vacuum down to 80 mbar (one-stage pumps), 9 mbar (two-stage pumps), 2 mbar (three-stage pumps) or 0.3 mbar (four-stage pumps).
These vacuum pumps are totally oil free and are the environmentally friendly alternative to water jet pumps as they do not consume water and, therefore, they do not produce contaminated waste water.
Diaphragm pumps can be used in a multitude of applications for vacuum generation in laboratories and process plants, such as vacuum filtration, vacuum degasing, vacuum impregnation, vacuum drying chambers, rotary evaporators, distillation, gel dryers, gas analytical systems and roughing of wide-range turbo molecular pumps
VACUUBRAND>REG> ROTA
Laboratory Rotary Vacuum Pumps
Rotary vacuum pumps are typically used when an ultimate vacuum in the 1 mbar to 10-3 mbar range is required. Rotary vacuum pumps are oil sealed, high performance, direct drive vacuum pumps. Most models come with various accessories such as separators, full-flow oil filters with maintenance indicator and oil mist filters.
Ideal for applications in chemistry laboratories, they offer a high vapour capacity for water and solvents and an active corrosion protection through an oil shut-off circuit, keeping the module under vacuum and stopping corrosives from entering the pump during non-operation.
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish