Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
80 pages of flow meters and hundreds of pages of flow accessories including pumps, fittings, tubing and process control. Request yours today.
Featured Products
Compact Gas Mass Flowmeters
gas mass flowmeter
  • 0 to 5 VDC and 4 to 20 mA analogue output
  • Optional totaliser available to display total accumulated flow
  • Flow rates up to 1000 sL/min with pressure drops up to 687 mbar
from $582.00 More details
Compact Gas Mass Flow Controllers
gas mass flow controllers
  • Precisely control flow rates with built-in valve
  • Control flow rates by either adjusting the local setpoint potentiometer or by supplying 0 to 5 VDC or 4 - 20 mA output signal
  • Flow rates up to 1000 sL/min with pressure drops up to 3.23 bar
from $882.00 More details
Cole-Parmer's Low Cost Flowmeters
low cost gas mass flowmeters
  • All units calibrated for air/nitrogen - calibration for other gases or gas mixtures available
  • Feature 0 to 5 VDC output signals for datalogging or controlling other instruments
  • Flow rates from 0 to 500 sL/min available
from $301.00 More details
Cole-Parmer's Low Cost Flow Controllers
Low cost gas mass flow controllers
  • Triple calibrated models available for use with air, helium and argon
  • Controllers with a display also feature a potentiometer for manual valve control
  • Flow rates of 0 to 10 sL/min
from $811.00 More details
C-P Catalogue > Flow Meter > Gas Mass Flowmeters & Controllers
Gas Mass Flow Meters and Gas Mass Flowmeter Controllers
Cole-Parmer gas mass flow meters and mass flow controllers can handle a wide range of gases including nitrogen, oxygen and argon. They are available in either aluminum or 316 stainless steel bodies. Cole-Parmer gas mass flow meters can measure gas flow from 0 sccm to 1000 sL/min with high accuracy models offering ±0.5% accuracy of reading. Our gas mass flow meter output options include mass flow display, totalizer display, 0 to 5 VDC or 4 to 20mA analogue signal. Order an analogue to RS-232 converter to allow data collection and analysis on your PC.
When measuring the flow rate of a compressible gas, changes to its pressure, temperature or density will result in changes to the volumetric reading. As mass readings are largely independent of pressure, temperature and density Cole-Parmer gas mass flow meters offer an ideal solution if you expect pressure or temperature variations in your system.
To find the right flow meter for your application use Cole-Parmer's on-line flow meter quick search. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs. Cole-Parmer offers both thermal dispersion and differential pressure gas mass flow meters.
Key Points for thermal dispersion gas mass flow meters
gas mass flowmeter technology
  • Inside a thermal gas mass flowmeter, the gas is split. Most goes through a bypass tube, while a fraction goes through a sensor tube containing two temperature coils.
  • Heat flux is introduced at two sections of the sensor tube by means of two wound coils.
  • As gas flows through the device, it carries heat from the upstream, to the downstream, coils.
  • The temperature differential, generates a proportional change in the resistance of the sensor windings.
  • Special circuits monitor the resistance change, which is proportional to mass flow, and calibrate it to give a digital readout of the flow.
  • The mass flowmeter is available with a built-in valve for flow-control applications. This allows for external control and the programming of a setpoint for a critical flowpoint. Most mass flowmeters also have an analogue or digital output signal to record the flowrate. The average mass flowmeter has an accuracy of ±1.5-2% of fullscale flow
Advantages for thermal dispersion gas mass flow meters
  • Ability (within limitations to ignore fluctuating and changing line temperatures and pressures
  • No moving parts
  • Measures the mass of gas, not volume, so it's very accurate
  • Accumulated flow can be logged using an optional totalizer
  • Inline meters available in a range of body material
  • Allows measurements of low flow rates
Disadvantages for thermal dispersion gas mass flow meters
  • Gas must be dry and free of particulates
  • Fairly slow response times
  • Mass flowmeters must be calibrated for a given gas or gas blend
Typical Application for thermal dispersion gas mass flow meters
  • Monitoring and controlling air flow during gas chromatography
  • Monitoring CO2 for food packaging
  • Gas delivery and control for fermenters and bioreactors
  • Leak testing
  • Hydrogen flow monitoring (e.g., in the utility industry)
  • Control of methane or argon to gas burners
  • Blending of air into dairy products
  • Regulating CO2 injected into bottles during beverage production
  • Nitrogen delivery and control for tank blanketing
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish