Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
80 pages of flow meters and hundreds of pages of flow accessories including pumps, fittings, tubing and process control. Request yours today.
Featured Products
Variable Area Flowmeters - Direct reading
Variable Area Flowmeters
  • Use with air sampling equipment, chromatography systems, gas analyzers, and water treatment systems
  • Bench mount with a tripod base
  • Machined from solid acrylic blocks, Integral metering tubes for precise readings
from $50.00 More details
65-mm Direct Reading Flowmeter
65-mm Direct Reading Flowmeter
  • Several options for valves and wetted materials.
  • Direct reading flowmeters for argon, CO2, helium, nitrogen, and oxygen
  • Fused ceramic millimeter scale & vertical tangential locator line ensures hairline accuracy in reading
from $80.00 More details
Thermal Gas Mass Flowmeter
Thermal Gas Mass Flowmeter
  • Calibrated for Air/N2
  • Available in variety of wetted materials - Anodized aluminum, brass, Viton®
  • Output - Linear 0 to 5 VDC, 4 to 20 mA, digital display
from $511.00 More details
Cole-Parmer Mass Flowmeters
Cole-Parmer Mass Flowmeters
  • Achieve accurate, ultrafast mass measurement and control
  • Measure any one of 16 standard gases through a simple key-pad setting
  • Units provide data on multiple fluid parameters: flow, pressure and temperature
from $1576.00 More details
Turbine Flowmeters/Totalizers
Turbine Flowmeters/Totalizers
  • Ideal low-cost solution for use in light manufacturing, pilot-plant, or quality control areas
  • NEMA enclosure-suitable for indoor/outdoor use in dusty, dirty and wet environments
  • Display flow rate and accumulated flow in GPM or LPM
from $425.00 More details
Magnetic Flowmeters
Krohne Magnetic Flowmeter
  • Designed specifically for the water and wastewater industry
  • Easy-to-install flow converter features a large, illuminated graphic display with intuitive programming
  • Designed for long-term reliability and durability, and are unaffected by contamination
from $2293.00 More details
Cole-Parmer Catalogue > Flow Meters and Flowmeter Controllers
Flow Meters and Flowmeter Controllers
Cole-Parmer offers an extensive range of flow meters and flow controllers for both liquid and gas applications. Cole-Parmer specializes in flow control of media in pipe lines less than 2" and offers flow metering solutions for measuring liquids flow from 0.0002 ml/min to over 4,000 LPM and for gases from 0.02 ml/min to over 500 m3/hour.
Cole-Parmer's flow meters come with a wide range of output options including set-point switches, analogue outputs 0(4)-20mA, 0-5 VDC, pulse signals and RS-232 options. Use Cole-Parmer's flow meter quick search to find the right flow meter for your application. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve you're application needs.
Flow Meter Selection Considerations
When selecting a flow meter consideration must be given to:
  • Flow measurement type - momentum (velocity), volumetric or mass flow measurement
  • Media - type of media (liquid, gas or slurry) and any special condition such as particulates in the media and viscosity of the media
  • Media conditions - pressure and temperature of media and whether media conditions are likely to remain constant or vary
  • Flow range - required flow range of media (min and max readings required)
  • Accuracy - required accuracy of the readings
  • Environmental considerations - special installation considerations such as hygienic installation, installing in to an ATEX zone or requiring tamper proof readings
For special or unique flow control applications
Use our helpful Flowmeter and Flow Meters Controller Application Table to narrow your selection. You will find information on application parameters like these:
  • Low-flow measurement/control (some types as low as 1 sccm air and 0.1 mL/min water, max of range)
  • High-flow measurement/control (some types as high as 65,000 LPM gas and 10,000 LPM liquid)
  • High pressure (34 bar and up)
  • High temperature (100°C and up)
  • High-accuracy measurement (error at or below 1%)
  • Aggressive or pure fluids
  • Fluids with heavy particulates or slurries
  • High-viscosity fluids
  • No wired power available
  • Intrinsically safe
  • Large pipe sizes (3" or larger)
  • Sanitary (meets 3A standards)
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams