Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 40 pages of filtration products including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Cole-Parmer UK Catalog > Filtration > Filtration Sizing
Filtration Sizing
Use this general guide to determine the optimal type of filtration and sizing for your particle sizes. Each type of filtration is designed to separate differing ranges of microparticles and particles:
  • Ultrafiltration is best with particles of roughly 10,000-1,000,000 MW;
  • Microfiltration is best with particle sizes of roughly 0.1-10im;
  • Macrofiltration is best with particle sizes greater than 50 im.
Type of filtration
Ultrafiltration
Microfiltration
Macrofiltration
Units
Dalton (MW)
Microns (µm)
50 µm or greater;
visible to
the human eye
Particle examples
and sizes
Trypsin (24,000)
Serum albumin (67,000)
Gamma globulin (169,000)
Viruses (600,000 to 30,000)
Endotoxins (500,000 to 12,000)
Mycoplasm (0.08 to 0.4)
Proteins (0.0005 to 0.5)
Fungi (0.7 to 3)
Yeasts (0.6 to 4)
Human red blood cells (3.5 to 8)
Bacteria (0.3 to 12)
Spores
Pollen
Human hair
Sands
Use Cole-Parmer's on-line catalogue to find and compare filtration specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of filtration experience to help solve your application needs.
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish