Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 40 pages of filtration materials and products including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Cole-Parmer Catalogue > Filtration > Filtration Materials
Filtration Materials
Types of filter membranes materials available at Cole-Parmer :
  • Cellulose Nitrate - most popular, used in analytical and laboratory filtrating. Material has excellent wetting properties and gives fastest flow rates with aqueous solutions
  • Cellulose Acetate - mixture of cellulose triacetate and diacetate which creates a strong membrane in both lateral and longitudinal directions. Additionally membrane has a low static charge, a very low aqueous extractability and good solvent resistance to low molecular weights alcohols
  • Nylon - strong membrane, inherently hydrophilic and compatible with a broad range of aqueous solutions including alcohols and solvents used in HPLC work
  • Polyethersulfone - membrane is hydrophilic and low protein binding. No external wetting agents required, resulting in low extractables. PES membranes generally offers fast flow rate and better chemical resistance than cellulose acetate membranes
  • Mixed Cellulose Ester - membrane provides a more uniform and smoother surface compared to pure nirocellulose membrane. This membrane is typically used to count or analyse particles contained in liquids or captured from aerosols.
  • PTFE - membrane is strong, highly porous and inert to most chemically aggressive solvents, strong acids and bases
Use Cole-Parmer's on-line catalogue to find and compare filtration specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of filtration experience to help solve your application needs.
Chemical Compatability For Membrane Selection
membrane compatability chart
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish