Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 40 pages of filtration housing products including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Cole-Parmer Catalogue > Filtration > Filtration Housing
Filtration Housing
Types of filter housings available at Cole-Parmer
Material of housing for filter holders available at Cole-Parmer include polysulphone (inexpensive), polypropylene, glass, SS, PTFE and other materials
Bottle-Top Filters;
Bottle Top Filters
  • Direct filtration into sterile glass media bottles
  • Vacuum port in middle juncture — vacuum allows for faster filtration due to pressure differential at filter membrane versus gravity feed
Nalgene Disposable S
Filter Units
  • Membrane holder, vacuum arm and receivers are complete system
  • Filter application for Cell Culture Media, Biological Fluids, Aqueous Solutions
Disposable In-Line Gas/Liquid Filters
In-line Filter Holders
  • Available with connections for tubing, pipes and syringes
  • Range of materials for suitability with applications such as liquids or gas filtration for acid, oil, high purity compressed gas and biological media
Millipore Microcon<r
Centrifuge Filters
  • More effective filtration due to extreme pressure at filtration membrane from centrifugal forces.
  • Advantages — highly reproducible, minimal sample loss
  • Applications — sample concentration, protein, peptide, DNA purification, buffer exchange, salt & radio label removal
Glass Fiber Prefilters with Final Nonsterile Cellu
Syringe Filters
  • Bi-directional filters
  • Available with luer slip and lock fit connections
  • Applications — HPLC, microbiological analysis, waste water treatment, sterilization of aqueous solutions, cryogenic fluids, macromolecules, protein binding
Glass Fiber Prefilters with Final Nonsterile Cellu
Vacuum filtration Holders and Manifolds
  • Connect 3-6 filter holders to one vacuum source. 2-3 way valves available to control vacuum or venting.
  • Application — ideal for testing of antibiotics and pharmaceuticals b bacteria retentive methods
Polypropylene bag fi
Bag Filters
  • High flow up to 150 LPM
  • Large solids — 1 to 800 µm
  • Pressure ratings up to 6.8 bar
  • Range of compatibility with organic solvents, animal fat, petroleum, vegetable oils, micro-organisms, alkalis, organic acids, oxidizing agents and mineral acids
  • Available with pressure gauge
Hayward <small><sup>�</sup></small>  single-
Heavy duty bag filter
  • High flow up to 830 LPM
  • Available with a wide range of bags for temperature, flow rate and filtration quality
  • Offer 98% efficiency at rated pore size
  • Easy bag change out
  • Applications — filtration of coolants, solvents, process water, ground water, oils for foods, lubrication and hydraulics
Double open end Slim
Cartridge style filters
  • Housing sizes up to 20" in metal and plastic designs for standard and high purity applications
  • Temperatures up to 157°C
  • Pressures up to 17 bar
  • Stacked systems — up to 20 double open end cartridges in one housing
Cartridges types
Double open end cartridges
  • Over 50 items with standard diameter of 2.75" for sediment, organic, lead, scale, and deionization treatment
  • Filtration down to 0.2 micro with flows up to 76 LPM
  • Includes variations that are autoclavable, biologically safe and FDA approved
High flow double open end cartridges
  • 4.5" diameter for sediment, organiscs, oil, iron and deionization treatment
  • Filtration down to 0.5 microns with flows up to 190 LPM
Single open end cartridges
  • Includes antimicrobial elements with filtration down to 0.1 micron
  • Cartridges meet USP, FDA and NSF criteria
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish