Help
Still have questions?
Expert Technical Service
Contact our experts
for FREE product application service
and support.
FREE 2009-2010 General Catalog
With over 30 pages of conductivity products and extensive sections in water and environmental quality including oxygen, pH, temperature and water quality, this catalogue offers thousands of solutions for your lab, process and field needs Request yours today.
Featured Products
Basic Handheld Conductivity Meter
handheld conductivity meter
  • Automatic / manual temperature compensation
  • Autoranging conductivity or TDS with temperature measurements
  • Selectable cell constant
from $343.00 More details
Benchtop Conductivity Meter
benchtop conductivity meter
  • Toggle easily between pH, conductivity, and TDS measurements
  • Dual display of pH, conductivity, or TDS plus temperature
  • Five-point calibration for pH and either conductivity or TDS
from $1019.00 More details
Pocket Conductivity Testers
Pocket Conductivity Testers
  • Dual-range measurements increase application range
  • Single or one point per range calibration
  • Replaceable sensor prolongs the life of your instrument
from $83.00 More details
Cole-Parmer Catalogue > Laboratory Equipment > Conductivity Meter & Measurement
Conductivity Meter & Measurement
Cole-Parmer UK offers a wide range of conductivity meters and conductivity controllers for a variety of environments and applications. Our low cost pocket conductivity testers and portable conductivity meters are ideal for field based conductivity measurement. Many models have an onboard memory to store and recall conductivity readings taken. For laboratory based applications, select from benchtop conductivity meters including leading brands such as Oakton, YSI, and Thermo Orion. Features of this style of conductivity meter can include a computer interface either by RS-232 or USB. Speak to our Applications Specialists for free technical support on conductivity measurement to help solve your application needs.
When considering features for a conductivity meter it is important to first understand how conductivity measurements are made. Conductivity is the ability of a material to conduct electric current. Determination of conductivity is achieved by the instrument by placing two plates in the sample, applying a potential across the plates and measuring the current that passes through the solution. Conductivity, the opposite of resistivity, is determined from the voltage and current values according to Ohm’s Law.
Conductivity Meter & Measurement Selection Considerations
When selecting a conductivity meter important features that need to be considered are :
  • Autoranging: Meter automatically selects the most accurate range for measurement. There is no need to manually change the range.
  • TDS Conversion Factor: When a solution does not have a similar ionic water content to natural water or salt water, then a TDS conversion factor is needed to automatically adjust the TDS readings
  • Temperature Compensation: A cell with built-in temperature sensor allows the meter to make adjustments to the conductivity or TDS reading based on changes in solution temperature
  • Adjustable temperature coefficients: Alcohols and pure water are affected by changes in temperature differently than typical samples. An adjustable temperature coefficient allows the user to compensate for the properties of the measured solution
  • Adjustable cell constant: Adjusts the measurement to reflect use of a cell with a constant other than K = 1.0. Wide range meters may except cells between K = 0.01 and 10
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish