Liquid Chromatography (LC) & Gas Chromatography (GC)
Cole-Parmer offers Liquid Chromatography and Gas Chromatography products for applications in the academic laboratory or industry.
Choose from a range of Gas Chromatography products including:
- Gas Chromatograph with Thermal Conductivity Detector (GC-TCD) for gas analysis or for simple mixtures. The TCD responds well to oxygen, nitrogen, water and other non-hydrocarbons with detection limits in the 100 ppm range
- Gas Chromatograph with Flame Ionization Detector (GC-FID) to detect hydrocarbons down to 1.0 nanogram. It responds linearly over the majority of the range
- Gas Chromatograph with a combination TCD and FID detector (GC-TCD/FID)connected in series allowing you to perform two analyses of the same sample at once
- Auto samplers, Gas generators, Purifiers and other GC accessories including maintenance kits, ferrules, injection port liners and stainless steel tubing
- Gas Chromatography columns (GC columns) and column packing accessories including our range of packed columns and the Thermo Scientific TRACE GC capillary columns
The Liquid Chromatography range consists of:
- High performance liquid chromotography systems (HPLC systems) including HPLC binary gradient systems, isocratic HPLC systems and automatic gradient HPLC systems
- LC accessories
- Low-pressure LC columns and fraction collectors
- LC columns
Thin Layer Chromatography products include:
Cole-Parmer also supplies Solid-phase extraction products including cartridges, columns, plates & wells and spin tips for micro extraction.
|
| |||||||||||||||||||||||||||||||||||||||
|
|
|||||||||||||||||||||||||||||||||||||||
|
|
||||||||||||||||||||||||||||||||||||||