Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
80 pages of flow meters and hundreds of pages of flow accessories including pumps, fittings, tubing and process control. Request yours today.
Featured Products
Variable Area Flowmeters
VA meter
  • Low cost acrylic body
  • Choice of direct reading meters, with or without control valve
  • Air: 0.04 LPM - 3,400 LPM Water: 5 ml/min - 75 LPM
from $55.00 More details
65-mm Direct Reading Flowmeter
65-mm Direct Reading Flowmeter
  • Several options for valves and wetted materials.
  • Direct reading flowmeters for argon, CO2, helium, nitrogen, and oxygen
  • Fused ceramic millimeter scale & vertical tangential locator line ensures hairline accuracy in reading
from $95.00 More details
Multi-tube Systems For Mixing Applications
multi flowmeter
  • Choice of direct reading meters, with or without control valve
  • Create your own multi tube or gas proportioner
  • Available for Air, Argon, Carbon Dioxide, Helium, Hydrogen, Oxygen and Water
from $666.00 More details
Cole-Parmer Catalogue > Flow Meters > Variable Area Flow Meters
Variable Area Flow Meters
Cole-Parmer offer an extensive range of variable area flow meters for both liquid and gas applications. Cole-Parmer's variable area flow meters are simple to install and offer cost effective solutions for applications requiring visual indication of flow or flow rate. Cole-Parmer offers variable area flow meters for measuring liquids flow from 0.0002 ml/min to over 400 LPM and for gases from 0.02 ml/min to over 700 m3/hour.
Cole-Parmer's variable area flow meters are available in a wide range of materials including low cost acrylic and specialist options such as PTFE for aggressive media or stainless steel for high temperature and pressure applications. Many of our variable area flowmeters come with a wide range of control and output options including control valves, set-point switches and analogue outputs.
To find the right flow meter for your application use Cole-Parmer's on-line flow meter quick search. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs.
Key Points for variable area flow meters
  • Variable area flow meters operate on the principle that the flow of the media will alter the position of a float in a tapered tube. The position of the float in the tapered tube is proportional to the volumetric flow rate
  • Typical accuracy of variable area flow meters is ±3% full scale
  • A variable area flow meter normally has a turn down ratio in the region of 10:1
  • A variable area flow meter normally requires vertical installation and a straight pipe run (minimum length of five times the diameter of the pipe) before the flow meters inlet
Advantages for variable area flow meters
  • Low cost
  • Simple to install
  • Easy to use and virtually maintenance free
  • Wide choice of materials including all PTFE for aggressive fluids
  • Many control options including control valves, set-point switches and analogue outputs
Disadvantages for variable area flow meters
  • Lower accuracy compared to many other flow meter technologies
  • Affected by pulsation
  • Sensitive to fluid changes such as viscosity, density and temperature
Typical Application for variable area flow meters
  • Liquid flow meter or gas flow meter where high accuracy is not required
  • Measuring water and gas flow in plants or labs
  • Monitoring chemical lines
  • Purging instrument air lines (i.e. lines that use a valved meter)
  • Monitoring filtration loading
  • Monitoring flow in material-blending applications (i.e. lines that use a valved meter)
  • Monitor makeup water for food & beverage plants
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish