Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Laboratory Equipment  > Laboratory Ovens  
Laboratory Oven and Lab Gravity Ovens
  More Information Buy
Baths Baths & Circulators Shop online
Centrifuges" Centrifuges Shop online
Furnaces" Furnaces Shop online
Refrigerators Refrigerators & Freezers Shop online
Ultrasonic Ultrasonic Cleaners Shop online
Cole Parmer UK offer a range of laboratory ovens and lab gravity ovens for a variety of heating applications. Cole-Parmer’s laboratory oven range can reach temperatures up to 300oC and capacities from 19 litres to 410 litres. Whether your application is for drying, ageing, softening or sterilising a variety of heating technologies are available. View our selection below or call our application specialists for free support for help you find a suitable laboratory oven to suit your needs.
Gravity convection laboratory ovens – provide low cost alternative to mechanical convection ovens when temperature uniformity is not essential to the process. Ideal for applications in which a powder or substance that needs to be heated or dried could be disturbed by a fan-generated airflow
Image 1
            Gravity Convection Lab Oven
            • Capacity range from 19 litres to 116 litres
            • Temperature range from ambient + 5oC to 300oC
            • Microprocessor controller with LED display and integrated 0 to 99-hr timer
            • Internal 304 stainless steel chamber has round edges for easy cleaning
            • Optional access ports can be added to the unit to allow connection of external measuring equipment
            • Temperature control accuracy ±0.3oC
            • Prices starting from £672.00
Mechanical convection laboratory ovens – utilizes a motorized fan to force air across heating elements and into the warning chamber. The fan allows for a more consistent distribution of air and is designed for applications in which good temperature uniformity or faster heatup/drying time are needed
Image 2
            Mechanical Convection Lab Oven
            • Capacity range from 19 litres to 116 litres
            • Temperature range from ambient + 5oC to 300oC
            • Microprocessor controller with LED display and integrated 0 to 99-hr timer
            • Powerful air turbine offers fast and uniform drying of large quantities of samples
            • Models available with option of adjusting air exchange rate from 0% to 100%
            • Features a microprocessor PID controller, digital timer, one ramp function and a LED temperature/time display
            • Prices starting from £864.00
Image 3
            Programmable Mechanical Convection Lab Oven
            • Capacity range from 53 litres to 240 litres
            • Temperature range from ambient + 5oC to 300oC
            • Microprocessor with two programs of ten sections each or alternatively 1 program with 20 sections
            • Program controller functions include delayed ON, delayed OFF, temperature dependent delayed OFF
            • Ventilation can be adjusted by means of a 55mm diameter rear exhaust duct with ventilation flap and front side ventilation
            • Communication software allow output to computer or printer
            • Prices starting from £1405.00
Safety ovens – are a mechanical convection oven that has a blow-out panel to protect the end user from injury in the event of an explosion within the oven. The safety oven is not explosion-proof and will not prevent volatiles from combusting. However, in the event of an explosion, the blow-out panel is designed to direct the force of the blast away from the user.
Image 4
            Safety Lab Oven
            • Extended temperature range up to 350oC
            • Capacity 116 litres
            • High air-exchange rates of 400L/min
            • Acousitic and visual alarms
            • Spring mounted safety glass panel with splinter protection
            • Fulfil all the safety regulations for handling substances containing solvents in accordance with EN1539
            • Replaceable fresh air filter improves particle-free, standard-compliant drying processes
            • Prices starting from £3474.00
Vacuum oven - is used when lack of atmosphere or inert atmosphere is desired. Because heating must occur in a vacuum where there is no convection of air, it is accomplished by placing the sample of the shelving in the unit so heat is conducted from the side walls to the sample via the metal shelf. These unit require a separate vacuum pump.
Image 5
            Vacuum Lab Oven
            • Microprocessor-controlled temperature regulation
            • Jacket heating system for excellent precision
            • Temperature range from ambient + 10oC to 200oC
            • Capacity range from 19 litres to 50 litres
            • Spring-loaded metal safety catch pens oven door slightly to prevent overpressure conditions
            • Port connects to a needle valve to gently regulate gas or air into the lab oven without disturbing the samples
            • Prices starting from £2960.00
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007