Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Industrial Mixers  > Static Mixers  
Static Mixers
  More Information Buy
laboratory Laboratory Mixers Shop online
industrial Industrial Mixers Shop online
handheld Handheld Mixers Shop online
mixer Accessories for Industrial Mixers Shop online
laboratory Accessories for Laboratory Mixers Shop online
Clear PVC schedule 40 Pipe Mixers
• 3/8'' NPT (M) connection
• Ideal for visual check of your mixing process
Pipe Mixers
• Mixer length 280 mm with 12 elements
from  Call for price   More details
316 Stainless Steel Pipe Mixers
• 1/2'' NPT (M) connection
• Mixer length 127 mm with 6 elements
stainless steel pipe mixers
• Mix fluids as you pump them through a pipe line
from  Call for price   More details
Kynar schedule 80 Pipe Mixers
• 1'' NPT (M) connection
• 380 mm length mixer with 12 elements
kynar pipe mixers
• Economical and long lasting in-line mixers
from  Call for price   More details
Disposable In-Line Mixers - Unmounted
• Element OD (Tubing ID) up to 1/2''
• Up to 254 mm length with 24 elements
disposable inline static mixers
• Economical
from  Call for price   More details
Cole Parmer UK offers a wide selection of in-line static mixers for use in flexible tubing and piping. Our range includes lengths up to 88 cm, materials including PVC, 316 Stainless Steel and Teflon and internal dimensions up to 5cm. To find the right static mixer for your application use Cole Parmer UK’s on-line search or alternatively contact our Application Specialists for free technical support and let us bring over 50 years of mixing experience to help solve your application needs.
Static Mixers let you mix fluids as you pump them through a pipe line. The fluid moves through the mixer in an alternating clockwise/counterclockwise motion to ensure homogeneous product. They are virtually maintenance free and need no spare parts. A in line static mixer is efficient and can withstand pressures up to 100 bar with the smallest sizes. The mixing elements inside the pipe and tube mixers are securely attached to the walls of the mixer so that internal pressure changes cannot collapse elements and stop flow.
Unmounted mixer
Disposable In-line Static Mixers
  • Ideal for your one-time mixing applications.
  • These disposable polyacetal mixers are available unmounted or mounted in polypropylene tubing. Adjust unmounted mixer lengths to get optimal mixing action – shorten by cutting or extend by using more than one mixer.
  • An unmounted static mixer can be used with any standard tubing which has the same ID as the mixing element OD.
  • A mounted static mixer can be used with any compression fitting with the same tubing OD.
Clear PVC schedule 40 pipe mixer
Pipe In-line Static Mixers
  • Install this static mixer directly into your process line to achieve a 100% mixed output – mixing elements are welded inside the pipe.
  • All mixers have threaded ends with a choice of NPT connections up to 2.”
  • Available in materials including PVC, Teflon and stainless steel
  • Clear PVC mixers to allow visual checks of your mixing process.
Sanitary Mixer, 1/2'
Sanitary In line Static Mixers
  • These are made of 316 stainless steel for low-or high-viscosity food processing.
  • All internal surfaces are ground and polished to a sanitary (#4) finish and have removable elements for cleaning and visual inspection.
  • Our blade design are best for low viscosity (<1,500 cp) fluids. The faster the liquid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.
  • Our helical design models are best for high viscosities (>1500 Cp) fluids and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise and ice cream.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007