Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Industrial Mixers & Mixing 
Industrial Mixers & Mixing
  More Information Buy
laboratory Laboratory Mixers Shop online
industrial Industrial Mixers Shop online
handheld Handheld Mixers Shop online
mixer Accessories for Industrial Mixers Shop online
laboratory Accessories for Laboratory Mixers Shop online
High torque, direct drive mixers
• Available in speeds up to 1800 rpm
• Motor types include air or electric driven
high torque industrial mixers
• Available in mounting options of clamps and bungs with lipseal
from  Call for price   More details
Heavy duty, gear drive mixers
• Ideal for mixing high viscosity materials or large batches of water like fluids
• 3 speed options available of up to 350 rpm
gear driven mixers
• Includes 316 SS shaft and impeller
from  Call for price   More details
Cole Parmer UK offers an extensive range of industrial mixers for a variety of mixing environments and applications including batch mixers, drum mixers and static mixers for use in the chemical processing, food, paint, pharmaceutical and water industries. Our range of industrial mixers has the ability to mix capacities up to 20,000 litres and viscosities up to 10,000 cps.
Cole Parmer UK’s industrial mixers are available with air or electric driven drives and with options for fixed or variable speed. They have a wide choice of mounting options including universal clamp for mounting directly to a stand or a tank rim and sanitary mixer fixings including tri-clamp flange fittings. Mixer shafts and impellers are available in stainless steel or PTFE coated for highly aggressive mixing applications.
Use Cole-Parmer’s on-line catalogue to find and compare mixer specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of mixing experience to help solve your application needs.
When considering which industrial mixer is best for your application, consideration has to be given to the following areas:
  • Liquid viscosity – when you apply a force to a liquid it will display a different characteristic. This viscosity characteristic allows you to estimate the power of the mixer you will need for your application
  • Container capacity – tank diameter, batch height and vessel shape
  • Process type – a mixer can be utilized for a range of different process including agitation, blending (the mixing of two or more liquids), dissolving of powder in to a liquid and solid suspension of particulates. The type of process will effect the positioning of the mixer, torque requirements, running speed and type of impeller used
  • Duty cycle – the time interval devoted to starting, running, stopping, and idling when a device is used intermittently
  • Power supply – electric or air driven mixers for areas where electricity is unsafe or impractical
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007