Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Laboratory Equipment  > Freeze Dryer & Drying Systems 
Freeze Dryer & Drying Systems
  More Information Buy
Baths Baths & Circulators Shop online
Centrifuges" Centrifuges Shop online
Furnaces" Furnaces Shop online
Refrigerators Refrigerators & Freezers Shop online
Ultrasonic Ultrasonic Cleaners Shop online
1L FreeZone Freeze Dryer
• Economical sample preservation - even with low-melting point solvents
• Convenient for small batch sizes
Freeze dryer
• Cascade cooling models lyophilize to -84° C
from  Call for price   More details
Clear Drying Chambers
• Available with 8 valved ports
• Available for ambient temperature use or heated to accelerate freeze
drying chamber
• Clear for easy visibility
from  Call for price   More details
Cole-Parmer UK offers a range of benchtop freeze dryers that are available with system capacities from 1L, 2.5L, 4.5L and 6.5L. Upon request with our technical staff we are able to offer console sizes up to 18L in capacities. Our range in freeze dryers include variations in refrigerator collectors that cool to -50°C and -84°C as well as teflon-coated models for more corrosive compounds. Our freeze dry systems are designed to handle the lyophilization needs of research and pilot plant laboratories, with the lower temperature collectors ideal for freeze drying samples with low eutectic temperature.
Freeze drying involves the removal of water or other solvents from a frozen product. Freeze dryers carry out three steps for this process – prefreezing, primary drying and finally secondary drying. Freeze drying has been used in a number of applications however it is most commonly used in the pharmaceutical industry, for example stabilization of living material such as microbial cultures, preservation of animal specimens and the concentration and recovery of reaction products. once freeze dried properly, products do not need refrigeration and items can be stored at ambient temperatures.
To complete your freeze drying systems, you will require the following components:
  • Freeze Dryer – Cole-Parmer’s catalogue range carries capacities up to 6L with temperatures of -54°C and -84°C, however larger capacities of up to 18L are available upon request.
  • Drying System Platforms - Depending on your application, Cole-Parmer’s freeze dryers offer a range of drying platforms. The drying platforms are the interfacing device that conducts solvent from the sample to the freeze dryer and may connect or hold sample containers.
  • Chamber platform
    Either accommodates sample shelving or provides flexible port connectivity for up to 12 or 16 sample flasks. Shelves maybe used for either bulk samples or bottles and are available with heaters to accelerate the freeze drying process.
    Manifold
    For parallel lyophilization using small containers
    Stoppering Tray Dryer
    For batches of small vessels or bulk sample on shelves. Vessels are then vacuum sealed in situ
  • Vacuum Pump – Also available chemical resistant models to enable freeze drying with more aggressive solvents
  • Sample containers - Sturdy, borosilicate glass vessels options include flasks, serum bottles, vials, and ampules
  • Add this option:
    • Shell Freezer for preliminary freezing of samples. Improves freeze drying efficiency by maximizing samle surface area in cylindrical fast free flasks. Available as stand-alone or integrated unit.
    Typical Application for freeze dryers
    • Freeze Drying Fish Samples for Volatile Substances
    • General Freeze Dry Application for Snake Venom
    • Freeze Drying Fungi
    • General Freeze Dry Application for Monitoring Soil Samples
    • Freeze Dry Applications for Monitoring Anti-Cancer Drugs
    • Freeze Drying Mango Powder
    Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
    Cole-Parmer Sites
    Cole-Parmer: United States
      Canada
      China
      United Kingdom
      India
    MasterFlex.com - Ordering and Technical Info
    FoodTechSource.com - Food Quality Site
    InnocalSolutions.com - Calibration and Certification
    Cole-Parmer
    Unit 3 River Brent Business Park
    Trumpers Way, Hanwell, London W7 2QA
    Telephone: 020 8574 7556
    Fax: 020 8574 7543
    E-Mail: sales@Coleparmer.co.uk
    Copyright © 2007