Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Tubing & Pipe Connecters, Fittings 
Fittings – Tubing Connectors and Pipe Fittings
  Buy Fittings Buy
barbed Barbed Fittings Shop online
Compression Compression Fittings Shop online
Threaded Threaded Fittings Shop online
Quick-Disconnect Quick-Disconnect Fittings & Couplings Shop online
Chromatography Chromatography Fittings Shop online
Luer Luer Fittings Shop online
Sanitary Sanitary Fittings Shop online
Cole-Parmer can provide quality tubing connectors and pipe fittings to help you set up your fluid handling system with no leaks, flow restrictions or costly downtime. Cole-Parmer’s tubing connector range includes barbed fittings for flexible tubing, compression fittings or pipe connectors for rigid tubing. Tubing connectors and pipe fittings are available in sizing to connect anything from microbore tubing up to pipes. We offer a variety of materials such as low cost polypropylene (PP) for general applications to PTFE for corrosive media, formulations that comply with FDA and USP regulations and for no extra charge we can offer certificates of fittings compliance our range in tubing connectors and pipe fittings.
Cole-Parmer provide you with the information you need to select an optimal fitting. We provide the basic physical property information including temperature and pressure rating for each material formulation. Our chemical resistance charts help determine the fitting material with best chemical compatibility to your process. Our selection includes:
Barbed Fittings
Slides into flexible tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations
      • Sizing: microbore – 2” ID
      • Available materials include: HDPE, Nylon, PP, Kynar, 316SS
Compression
Have a nut that compresses rigid tubing against the fitting body for high pressure applications
      • Sizing: 1/16” to ¾” OD
      • Available materials include: HDPE, Nylon, PP, Kynar, 316SS
Pipe Fittings
Threaded in NPT (National Pipe Taper), UNF (Unified Find Thread) or BSP (British Standard
Pipe) sizes for permanent, high-pressure pipe connections
      • Sizing: 1/8” to 1” NPT and BSP
      • Available materials include: PP, Nylon, PTFE, 316 SS
Valved fittings
Allow you to make and break tubing connections without separating tubing from fitting. Available
with barbed, pipe compression, and push-to-connect connections
      • Available materials include: PP, brass, PTFE, Stainless Steel
      • Available with compression, threaded and hose barb connections, as well as valve or straight-through couplings
Multiport connector; tee; tube OD, 1 to 4 mm
Type of compression fitting designed for HPLC and other with high working pressures (from
68 to 680 bar)
      • Available in straight connectors or multiport, up to 8 ways
      • Material include PTFE
PVC Luer Adapters
Designed to work with micro tubing and luer-type syringes. Standard-sizing connecting end make
luer fittings, stopcocks and manifold completely interchangeable
      • Ideal for syringe or small tubing applications from 1/16” to ¼”
      • Stopcocks and manifolds available
Sanitary
Designed for contamination free tubing connections. Most of these autoclavable fittings meet FDA
and USDA requirements and 3A sanitary standards
Ideal for syringe or small tubing applications from 1/16” to ¼”
      • Sizing: 1/2" to 2" clamps
      • Available material includes PP, PVDF, PFA and 316 stainless steel and 304 stainless steel
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007