Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Filtration > Filtration Sizing  
Filtration Sizing
  More Information Buy
bottle Bottle Top/Filter Units Shop online
in In-Line Filters/Capsules/Hollow Fiber Shop online
filter Filter Paper/Filter Membranes Shop online
syringe Syringe Filters Shop online
vacuum Vacuum/Pressure Filters Shop online
cartridge Cartridge Style Filters for Liquids Shop online
bag Bag Style Filters for Liquids Shop online
ultrapure Filters for Ultrapure liquids and gases Shop online
Use this general guide to determine the optimal type of filtration and sizing for your particle sizes. Each type of filtration is designed to separate differing ranges of microparticles and particles:
  • ultrafiltration is best with particles of roughly 10,000-1,000,000 MW; microfiltration is best with particle sizes of roughly 0.1-10ìm;
  • macrofiltration is best with particle sizes greater than 50 ìm.
Type of filtration
Ultrafiltration
Microfiltration
Macrofiltration
Units
Dalton (MW)
Microns (µm)
50 µm or greater;
visible to
the human eye
Particle examples
and sizes
Trypsin (24,000)
Serum albumin (67,000)
Gamma globulin (169,000)
Viruses (600,000 to 30,000)
Endotoxins (500,000 to 12,000)
Mycoplasm (0.08 to 0.4)
Proteins (0.0005 to 0.5)
Fungi (0.7 to 3)
Yeasts (0.6 to 4)
Human red blood cells (3.5 to 8)
Bacteria (0.3 to 12)
Spores
Pollen
Human hair
Sands
Use Cole Parmer UK’s on-line catalogue to find and compare filtration specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of filtration experience to help solve your application needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007