Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Filtration > Filtration Materials 
Filtration Materials
  More Information Buy
bottle Bottle Top/Filter Units Shop online
in In-Line Filters/Capsules/Hollow Fiber Shop online
filter Filter Paper/Filter Membranes Shop online
syringe Syringe Filters Shop online
vacuum Vacuum/Pressure Filters Shop online
cartridge Cartridge Style Filters for Liquids Shop online
bag Bag Style Filters for Liquids Shop online
ultrapure Filters for Ultrapure liquids and gases Shop online
Chemical Compatability For Membrane Selection
membrane compatability chart
Types of filter membranes materials available at Cole-Parmer :
  • Cellulose Nitrate – most popular, used in analytical and laboratory filtrating. Material has excellent wetting properties and gives fastest flow rates with aqueous solutions
  • Cellulose Acetate – mixture of cellulose triacetate and diacetate which creates a strong membrane in both lateral and longitudinal directions. Additionally membrane has a low static charge, a very low aqueous extractability and good solvent resistance to low molecular weights alcohols
  • Nylon – strong membrane, inherently hydrophilic and compatible with a broad range of aqueous solutions including alcohols and solvents used in HPLC work
  • Polyethersulfone – membrane is hydrophilic and low protein binding. No external wetting agents required, resulting in low extractables. PES membranes generally offers fast flow rate and better chemical resistance than cellulose acetate membranes
  • Mixed Cellulose Ester – membrane provides a more uniform and smoother surface compared to pure nirocellulose membrane. This membrane is typically used to count or analyse particles contained in liquids or captured from aerosols.
  • PTFE – membrane is strong, highly porous and inert to most chemically aggressive solvents, strong acids and bases
Use Cole-Parmer’s on-line catalogue to find and compare filtration specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of filtration experience to help solve your application needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007