Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Filtration > Filtration Housing  
Filtration Housing
  More Information Buy
bottle Bottle Top/Filter Units Shop online
in In-Line Filters/Capsules/Hollow Fiber Shop online
filter Filter Paper/Filter Membranes Shop online
syringe Syringe Filters Shop online
vacuum Vacuum/Pressure Filters Shop online
cartridge Cartridge Style Filters for Liquids Shop online
bag Bag Style Filters for Liquids Shop online
ultrapure Filters for Ultrapure liquids and gases Shop online
Types of filter housings available at Cole-Parmer
Material of housing for filter holders available at Cole-Parmer include polysulphone (inexpensive), polypropylene, glass, SS, PTFE and other materials
Bottle-Top Filters;
Bottle Top Filters
  • Direct filtration into sterile glass media bottles
  • Vacuum port in middle juncture – vacuum allows for faster filtration due to pressure differential at filter membrane versus gravity feed
Nalgene Disposable S
Filter Units
  • Membrane holder, vacuum arm and receivers are complete system
  • Filter application for Cell Culture Media, Biological Fluids, Aqueous Solutions
Disposable In-Line Gas/Liquid Filters
In-line Filter Holders
  • Available with connections for tubing, pipes and syringes
  • Range of materials for suitability with applications such as liquids or gas filtration for acid, oil, high purity compressed gas and biological media
Millipore Microcon<r
Centrifuge Filters
  • More effective filtration due to extreme pressure at filtration membrane from centrifugal forces.
  • Advantages – highly reproducible, minimal sample loss
  • Applications – sample concentration, protein, peptide, DNA purification, buffer exchange, salt & radio label removal
Glass Fiber Prefilters with Final Nonsterile Cellu
Syringe Filters
  • Bi-directional filters
  • Available with luer slip and lock fit connections
  • Applications – HPLC, microbiological analysis, waste water treatment, sterilization of aqueous solutions, cryogenic fluids, macromolecules, protein binding
Glass Fiber Prefilters with Final Nonsterile Cellu
Vacuum filtration Holders and Manifolds
  • Connect 3-6 filter holders to one vacuum source. 2-3 way valves available to control vacuum or venting.
  • Application – ideal for testing of antibiotics and pharmaceuticals b bacteria retentive methods
Polypropylene bag fi
Bag Filters
  • High flow – 150 LPM
  • Large solids – 1 to 800 um
  • Pressure ratings up 6.8 bar
  • Range of compatibility with organic solvents, animal fat, petroleum, vegetable oils, micro-organisms, alkalis, organic acids, oxidizing agents and mineral acids
  • Available with pressure gauge
Hayward <small><sup>®</sup></small>  single-
Heavy duty bag filter
  • High flow – 830 LPM
  • Available with a wide range of bags for temperature, flow rate and filtration quality
  • Offer 98% efficiency at rated pore size
  • Easy bag change out
  • Applications – filtration of coolants, solvents, process water, ground water, oils for foods, lubrication and hydraulics
Double open end Slim
Cartridge style filters
  • Housing sizes up to 2” in metal and plastic designs for standard and high purity applications
  • Temperatures up to 157oC
  • Pressures up to 17 bar
  • 20” stacked systems
Cartridges types
Double open end cartridges
  • Over 50 items with standard diameter of 2 ¾” for sediment, organic, lead, scale, and deionization treatment
  • Filtration down to 0.2 micro with flows up to 76 LPM
  • Includes variations that are autoclavable, biologically safe and FDA approved
High flow double open end cartridges
  • 4 ½” diameter for sediment, organiscs, oil, iron and deionization treatment
  • Filtration down to 0.5 microns with flows up to 190 LPM
Single open end cartridges
  • Includes antimicrobial elements with filtration down to 0.1 micron
  • Cartridges meet USP, FDA and NSF criteria
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007