Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Laboratory Filtration & Filtering 
Laboratory Filtration & Filtering
  More Information Buy
bottle Bottle Top/Filter Units Shop online
in In-Line Filters/Capsules/Hollow Fiber Shop online
filter Filter Paper/Filter Membranes Shop online
syringe Syringe Filters Shop online
vacuum Vacuum/Pressure Filters Shop online
cartridge Cartridge Style Filters for Liquids Shop online
bag Bag Style Filters for Liquids Shop online
ultrapure Filters for Ultrapure liquids and gases Shop online
Laboratory Filtration & Filtering is a simple technique for purification of liquids and gases. The basic principle of operation is that the fluid passes through a porous structure to remove solid particulates. The pore size of the filter media determines which particles are allowed to pass. Cole Parmer UK offers a wide selection in filter media as well as housing for the filter to best suit a variety of applications in both the laboratory and process applications.
Cole Parmer UK’s range of filtration products includes filter membranes which are able to retain particles sizes from 0.1 microns, filter material including nylon to be used with alcohols and solvents along with PTFE for the most chemically aggressive solutions. Our filter housings are available in a range of styles including in-line filters, vacuum and cartridge systems. They are able to handles small volumes as low as 1ml and flow rates up to 830 LPM. When selecting a filtration system for your application consideration has to be given to :
  • Filter membrane size - the size of the particle to be retained or filtered. Membranes will retain all particles equal to, and larger than their designated pore size
  • the filter chemical compatibility to the membrane with the liquid or gas to be filtered. Consider the chemical resistance properties of all the parts that will contact the filtrate
  • Filter housing style in terms of temperature, flow rate, throughput and sterilization needs
Use Cole Parmer UK’s on-line catalogue to find and compare filtration specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of filtration experience to help solve your application needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007