Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Laboratory Equipment > Chromatography  > Conductivity 
HPLC
  More Information Buy
gas Gas Chromatography Shop online
chromatography Chromatography Fittings Shop online
liquid Liquid Chromatography Shop online
thing Thin Layer Chromatography Shop online
Cole-Parmer HPLC Systems
• Models available with up to pressures of 5000 PSI
• Flow rate 0.01 to 9.99 ml/min
HPLC system
• Display shows absorbance, wavelength and pressure
from  £5667.00   More details
Cole Parmer UK offers a range of High performance liquid chromatography (HPLC) solutions for routine industrial process monitoring, education and quality control (QC). Our HPLC systems are suitable for both analytical and preparative separation methods.
HPLC is a common laboratory technique used for liquid separation of mixtures. Separation is achieved by using a liquid mobile phase to separate the components of the sample. The components are dissolved in a solvent and forced through a chromatography column packed with particulates or porous layers under high pressure. As the sample is resolved into its components they are then passed through a detector and a chromatograph is generated.
Cole-Parmer’s HPLC systems come with variable wavelength (UV and visible) detectors and offer a choice of Isocratic or Binary Gradient HPLC pump systems.
  • Isocratic HPLC systems are used for routine separation and analysis and work by using a constant mobile phase composition to elute compounds.
  • Cole-Parmer’s Binary Gradient HPLC systems include a second solvent pump that varies the strength of the organic solvent to elute different compounds. The system also comes with PC software for gradient and other parameter setups, data acquisition and system control making it ideal for most lab applications.
When selecting your Chromatography and HPLC equipment, consideration has to be given to a range of factors. View our online catalogue to see our full selection of products or contact our Application Specialist to help you find the right product for your needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007