United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers, Blenders, and Stirrers
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Refrigerant Gas Leak Detectors
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Browse all product categories
Alphabetically (A-Z)
Category Index
Industry/Discipline
A
B
C
D
E
F
G
H
I
J
K
L
M
N
O
P
R
S
T
U
V
W
A
Accessories for pH Meters and Ion Meters
Accessories for Spectroscopy
Actuators
Adhesives and Epoxies
Advanced Light Meter
Air Compressors
Air Sampling
Air Sampling and Gas Analysis
Airflow Measurement
Alarms
Analytical Balances
Animal Research Products
Apparel
Arc Flash Products
Atomic Absorption
Automation Control
B
Bags
Balance and Scale Accessories
Balances & Scales
Bath & Circulator Accessories
Baths & Circulators
Beakers
Benches and Tables
Biofuels
Biohazard Bags and Biosafety Disposal
Blenders and Paddle Blenders
Block Heaters
Books & Reference Materials
Bottles
Bowls
Brushes
Buckets
Buffers, Storage Solutions, and Standards
Burettes (Burets)
Burners
C
Cable Ties
Calculators
Calibration
Calibration Equipment
Calibration Masses
Calibration Services
Calibrators
Calipers
Cameras and Video Microscopy Equipment
Carboys
Carts, Mobile Cabinets, and Mobile Workstations
Cell Biology
Centrifugal Separation
Centrifuges
Chairs and Stools
Chemical Oxygen Demand Testing
Chemical Property Testing
Chloride Analyzers
Chromatography Products
Circulating Water Baths
Clamps and Supports
Clean Room
Clean Room Swabs
Clean Room Wipes
Cleaner/Degreasers and Contact Cleaners
Cleaners
Cleaning & Janitorial Supplies
Cleaning Brushes
Cleaning Solutions
Cleanroom and Static Control Gloves
Cleanroom Furniture
Color & Gloss Meters
Colorimeters
Colorimetry Reagents
Compound Microscopes
Conductivity/Resistivity/TDS Meters and Testers
Confined Space Gas Detectors
Containers
Cooling Equipment
Critical Environment Apparel
Crucibles
Cryogenics
D
Data Acquisition
Dataloggers
Density/Specific Gravity
Desiccators
Detergents
Dialysis
Dishes
Dispensers
Dispensing Pumps
Dispersion Mixers and Accessories
Dissolved Oxygen (DO) Probes and Sensors
Dissolved Oxygen Benchtop Meters
Dissolved Oxygen Handheld Meters
Dissolved Oxygen Test Kits
Document and Waste Containers
Drum and Hand Pumps
Drum Heaters
Dye Tracers
E
Electrical
Electrical Safety
Electrodes
Electrophoresis Products
Electroporation
Enpac Cylinder Dolly
Environmental Chambers
Environmental Testing
Ergonomics
Evaporators
Extraction Apparatus
Eyewash
Eyewear and Safety Glasses
F
Fermentation
Fiberscopes and Borescopes
Filling Pumps
Film and Foil
Filtration
First Aid
Fittings
Flame Photometers
Flasks
Floor Mats and Matting
Flow, Level & Valves
Flowmeters
Fluid & Liquid Handling
Fluid Bed Dryers
Fluorometers
Food Quality & Testing
Force & Torque
Forensics
Freeze Dryers
Freezers - Chest/Cabinet
Freezers - Upright freezers
Fume Extractors
Fume Hoods
Funnels
Furnaces
Furniture & Fume Hoods
G
Gaskets
Gauge guards, Seals, and Accessories
Gauges
Gear Pump Systems
Gear Pumps
Genomics
Glassware
Glassware Washers
Glove Boxes
Gloves
Graduated Cylinders
H
Hammers
Hand & Platform Trucks
Hand Tools
Handheld Mixers
Handlebar Pail and Drum Air Mixers
Health and Safety
Heat Guns
Heat Sealer
Heat Stress Monitors
Heated Water Baths
Heating Equipment
Heating Mantles
Heating Tapes, Cords, Blankets
Heating Torches
High Throughput Screening
Histology
Homogenizers
Hose
Hot Plates
Humidifiers/Dehumidifiers
Humidity Indicators (Dial)
Humidity Indicators (Digital)
Humidity Loggers and Recorders
Humidity Meters (Thermohygrometers Meters)
Humidity Pens (Thermohygrometer Pen-style)
Humidity Psychrometers / Mason Hygrometers
Humidity Switches / Panel Mount Controllers / Alarms/Monitors
Humidity Transmitters
Hydrometers
I
Ice Buckets and Ice Pans
Ice Machines
IKA Magic LAB Mixing System
Illuminated Magnifiers
Immersion Heaters
Immunochemistry
Immunology
Incubators
Industrial Mixers
Industrial Process
Infrared (IR) Spectroscopy
InnoCal
Peristaltic Pump Drive RPM Calibration
InnoCal
Pump Calibration and Repair Services
Inspection / Pick-up Tools
Instrument Calibration
Instrument Cases
Ismatec® Pumps
J
Junction Boxes
K
Knives
Konica Illuminance Meters
L
Lab Supplies & Consumables
Label Printers and Labels
Laboratory & Utility Water Baths
Laboratory Equipment
Laboratory Mixers
Laboratory Research
Labware
Ladles and Dippers
Laser Products
Level Control
Life Science
Light Meter with Analog Output
Light Meters
Lighting
Lighting and Illumination
Lightnin
Mixers and Accessories
Lockout/Tagout Product
Luer Fittings and Adapters
Luer Manifolds
Luer Stopcocks
Luer Valves and Assemblies
LUX Light Meter
M
Magnifiers
Manometers
Manostat Peristaltic Pumps
Masterflex
Puri-Flex
Microbore - Autoanalysis Tubing
Masterflex B/T Pump Tubing
Masterflex C/L Pump Tubing
Masterflex C/L Tubing Pumps
Masterflex Chem Durance
Tubing
Masterflex I/P Pump Tubing
Masterflex L/S Pump Tubing
Masterflex
Self-Contained Composite Sampler
Masterflex
Tubing Formulations
Masterflex® B/T® Tubing Pumps
Masterflex® I/P® Tubing Pumps
Masterflex® L/S® Tubing Pumps
Masterflex® Peristaltic Pumps
Masterflex® Pump Head Adapters
Masterflex® Pump Heads
Masterflex® Pump Tubing
Masterflex® Tubing and Pump Accessories
Material Storage & Handling
Material Testing & Physical Testing
Mats
Mattings and Liners
Measuring Tools
Mechanical Balances
Melting Point
Metal Detectors
Metering Pumps
Microbalances
Microbiology
Microfluidic Pumps
Microfluidics
Micrometers
Microscope Accessories
Microscope/Telescopes (Dual-Purpose)
Microscopes & Magnification
Midshaft Turbine Propeller
Mills
Mixers and Blenders
Moisture Balances
Moisture Meters
Moisture Water Activity Systems
Molecular Biology
Mop Handles
Mops
Mortars & Pestles
Motors
Multiparameter Meters
N
Natural Resources and Mining
Notebooks
Nuclear Magnetic Resonance (NMR) Spectroscopy
O
Orbital Motion Heated Bath
Ovens
P
Pail Mixer with Rim Clamp
Pails
Panel Meters
PCR (Polymerase Chain Reaction)
Pelican Watertight Instrument Cases
Pens and Markers
Peristaltic Pumps
Personal Protective Equipment (PPE)
pH Measurement
pH Meters, Ion Meters and ISFET Meters
pH Test Strips, pH Indicators, and Water Quality Testing Kits
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pharmaceutical Industry Products
Physical Property Testing
Pipettes and Pipettors
Piping
Plastic Storage Bins & Containers
Pliers
Pocket Testers
Polarimeters
Portable Balances
Positive Displacement Pumps
Powder Mixers
Precision Indicators
Presses
Pressure & Vacuum Controllers
Pressure & Vacuum Instruments
Pressure I/P Transducers
Pressure Regulators - Industrial
Pressure Regulators for Gases
Pressure Transmitters and Pressure Transducers
Process Controllers
Process Equipment & Instrumentation
Process Equipment, Process Instruments & Process Controllers
Proteomics
Pump Accessories
Pump Systems
Pumps
R
Racks
Radiation Detectors
Radios
Recirculators, Chillers, Immersion Chillers and Coolers
Recorders
Recorders & Data Acquisition
Refractometers
Refrigerant Gas Leak Detectors
Refrigerator and Freezer Combination
Refrigerators
Relative Humidity
Respirators and Masks
S
Safety Apparel
Safety Light Curtains
Safety Products
Safety Products for Life Science
Safety Shields
Safety Signs & Labels
Safety Software
Sample Containers
Samplers
Sanitary and Sterile Products
Sanitary Mixers
Sanitary Scrub Brushes
Sanitary Transducers, Gauges, and In-Line Seals
Scales
Scoops and Spoons
Screwdrivers
Self-Healing Cutting Mats
Shakers
Shaking Water Baths
Shelving
Shippers
Shop by Industry
Shop Microscopes
Sieves
Silicone Spray
Skincare
Soldering Tools
Sound Meters
Spatulas & Stir Rods
Specialty Heaters
Specialty Microscopes
Specialty spectrophotometers
Spectroscopy and Spectrophotometers
Spill Control
Spray Bottles
Stainless Steel Scrub Pads
Stainless Steelware
Static (In-line) Mixers
Stationary
Stationary and Portable Vises
Steam Generators
Steam Handling
Stem Cell Research
Stereo and Stereozoom Microscopes
Sterilizers
Stir Bars
Stirrers
Stools & Ladders
Stoppers
Storage and Handling
Storage and Utility Cabinets
Surface Testing
Switches
Synthesis Systems
Syringes
T
Tachometers/Stroboscopes
Tanks and Drums
Tanks for Baths and Circulators
Tapes
Temperature & Humidity Dataloggers
Temperature Bimetal Thermometers
Temperature Chart Recorders
Temperature Controllers
Temperature Dataloggers
Temperature Digital Thermometers
Temperature Glass Thermometers
Temperature Infrared Thermal Imagers
Temperature Infrared Thermometers / Sensors
Temperature Labels and Crayons
Temperature Probes and Accessories
Temperature RTD Instruments and Probes
Temperature Thermistor Instruments and Probes
Temperature Thermocouple Instruments and Probes
Temperature, Humidity and Moisture Products
Tensiometers
Test and Measurement Equipment
Test Tubes
Timers
Tissue and Cell Culture
Titrators
Tongs and Tweezers
Tool Kits
Tools
Toploading Balances
Trays
Tubing
Tubing, Fittings, and Hose
Turbidity Meters and Turbidity Testing Products
Tweezer Set
U
Ultrasonic Cleaners
Ultrasonic Processors
Ultraviolet Equipment
V
Vacuum Controllers, Gauges, and Systems
Vacuum Pumps
Vacuums
Valves
Vials
Vibration Measurement
Viscometers
Visible/UV-Visible Spectroscopy
W
Wall Mount Enclosures, Cabinets, & Racks
Water Pressure Loggers
Water Purification Systems and Accessories
Wave Biotech Hot Lips
Tube Sealer
Weather Testing Equipment
Western Blotting
Wide-Field Microscopes
Workstations
Worktables and Trucks
Wrenches
Free Catalogs
2011/2012
Cole-Parmer General Catalog
Request your FREE copy
of the catalog that has become an industry resource today!
Special Offers
Check out our
Special Offers
on a variety of products.
Technical Resources
Learn
from our comprehensive
knowledge base.
Over 1000 files
Expert Service
Customer satisfaction
on recommended products.
Full list of services
View Popular Product Categories:
Mass Flowmeters
|
Tissue Culture
|
Vacuum Pumps
|
Pressure Connectors
|
Radiation Detectors
|
Pipette Tips
View Top Selling Categories:
Peristaltic Pump
|
Laboratory Equipment
|
Chemical Cabinets
|
Test Tubes
|
Microbore Tubing
|
Ultrasonic Cleaners
View Featured Categories:
Liquid Nitrogen Dewar
|
Centrifugal Pumps
|
Centrifuge Tubes
|
Temperature Humidity Meter
|
Petri Dish