Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Laboratory Equipment  > Balances > Ohaus balances 
Ohaus Balance, Ohaus Scale and Wweighing from
  More Information Buy
Baths Baths & Circulators Shop online
Centrifuges" Centrifuges Shop online
Furnaces" Furnaces Shop online
Refrigerators Refrigerators & Freezers Shop online
Ultrasonic Ultrasonic Cleaners Shop online
Balances" Balances Shop online
Ohaus balances have served the laboratory, education and industrial sector for over 90 years. Cole Parmer UK offers a wide selection of the Ohaus balances ranging from portable scales, analytical balances, toploading balances, moisture balances and industrial scales. Ohaus supply reliable, precise and affordable balances to suit all your weighing needs.
Ohaus portable balances are ideal for use in light industrial, field or office uses. The analytical and toploading range from Ohaus includes features such as internal or external calibration, computer interface and draft-shield. Ohaus industrial scales have a capacity of up to 230kg and are available with NTEP approval which means they are legal for trade balances.
image1
Handheld Ohaus Balance
  • Capacity up to 320g
  • Readability to 0.1g (1 decimal place)
  • Convenient, compact and portable weighing
  • Battery Operated
  • Automatic shutoff
  • Starting price - £77.00
image2
Compact Ohaus scales
  • Capacity up to 5000g (5kg)
  • Readability to 0.1g (1 decimal place)
  • Digital keypad calibration with external weight
  • Stability Indicator
  • Starting price - £50.00
image3
Ohaus Scout Pro Portable Balance
  • Capacity up to 6000g
  • Readability to 0.01g (2 decimal places)
  • High contrast LCD display
  • Built in security bracket
  • Parts counting with auto optimization
  • Starting price - £170.00
image3
Adventurer SL Analytical Balance And Toploading Balances
  • Capacity up to 8100g for toploading balances and 210g for analytical balances
  • Readability to 0.001g for toploading balances and 0.1 mg for analytical balances
  • Easy-access, frameless draft shield with removable doors on analytical and high precision toploading models
  • Two-button keypad with dedicated yes/no functions simplifies balance use
  • Integrated security bracket
  • Starting price - £395.00
image3
Ohaus Adventurer Pro Analytical And Toploading Balances
  • Capacity up to 8100g for toploading balances and 260g for analytical balances
  • Readability to 0.001g for toploading balances and 0.0001g for analytical balances
  • Internal or external mass calibration
  • Built in bidirectional RS232, allowing connection to a PC or a printer
  • Easy access, frameless draft shield with removable doors
  • Starting price - £405.00
Ohaus MB 35 Moisture
MB-Series Moisture Analyzers
  • Capacity up to 40g
  • Readability up to 0.01%/0.001g
  • Fast halogen heating element provides uniform infrared heating and durability for extended use
  • Superior analytical weight performance
  • Standard RS-232 port for easy communication
  • Starting price £1785.00
image3
Ohaus Champ Bench Scales
  • Capacity up to 250 kg
  • Readability to 0.001kg
  • Features adjustable averaging levels and parts counting
  • Stainless steel wash down scales
  • IP 65 water tight
  • Starting price - £595.00
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007