Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Laboratory Equipment  > Balances > Analytical Balances 
Analytical Balances and Weighing Balance
  More Information Buy
Baths Baths & Circulators Shop online
Centrifuges" Centrifuges Shop online
Furnaces" Furnaces Shop online
Refrigerators Refrigerators & Freezers Shop online
Ultrasonic Ultrasonic Cleaners Shop online
Balances" Balances Shop online
For high precision applications where readability of weight is essential, Cole-Parmer UK is the leading distributors of analytical balances. Analytical balance models are available with readabilities of up to 0.01mg (5 decimal places) making them ideal for universities, pharmaceutical and high precision manufacturing applications. Cole-Parmer is able to offer balances with capacities up to 320mg with ranges from leading brands including – Cole-Parmer Symmetry, Ohaus, Mettler Toledo, A&D Engineering, Sartorius, Acculab and Adam equipment.
Our range in laboratory analytical balances includes a variety of features to suit your application needs. Models are available with internal or external calibration, built in RS-232 to allow connectivity to computers or printers and stability filters to compensate from vibrations, drafts or temperature changes. To ensure high accuracy weighing our analytical balances are available with a see-through enclosure to prevent dust, air or any other external factors to affect the reading of the balances.
To find the right analytical balance for your application use Cole-Parmer’s on-line search to help you shop by specification or by brand. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of weighing experience to help solve your application needs.
Acculab® Analytical
Acculab ALC Precision Analytical Balance
  • Capacities up to 210g
  • Readability up to 0.0001g (0.1mg)
  • Repeatability of ± 0.0001g (0.1mg)
  • Prices from £930.00
  • Built in bidirectional RS-232 interface
  • Digital filtering and fast stabilization
Adam PW Analytical B
Adam Equipment PW Precision Analytical Balance
  • Capacities up to 250g
  • Readability up to 0.0001g (0.1mg)
  • Repeatability of ± 0.0002g (0.2mg)
  • Prices from £750.00
  • GLP-compliant printouts via RS-232
  • Date and time stored in memory
120g
AND Analytical Balance
  • Capacities up to 250g
  • Readability up to 0.00001g (0.01mg)
  • Repeatability of ± 0.00001g (0.01mg)
  • Prices from £1195.00
  • Digital tare let you tare via computer or manually
  • Antistatic coating on glass to minimize static effects
Cole-Parmer<logo> Sy
Cole-Parmer Symmetry PA Precision Analytical Balance
  • Capacities up to 220g
  • Readability up to 0.0001g (0.1mg)
  • Repeatability of ± 0.0002g (0.2mg)
  • Prices from £737.00
  • Bidirectional RS-232 interface
  • Stabilization time of 2 to 4 seconds
Analytical Balance,
Mettler Toledo Analytical Balance
  • Capacity up to 320g
  • Readability up to 0.00001g (0.01mg)
  • Repeatability up to ±0.00003g (0.03mg)
  • Prices from £1215.00
  • Connect to peripheral devices via RS-232, Ethernet, or Bluetooth wireless technology
  • Dismountable draft shielding for easy cleaning
Ohaus <small><sup>®</sup></small>  Adventure
Ohaus Analytical Balance
  • Capacity up to 260g
  • Readability up to 0.0001g (0.1mg)
  • Repeatability up to ±0.0001g (0.1mg)
  • Prices from £795
  • RS-232 allows computer/printer connection to record GLP data including time, date, balance ID and user ID
  • User selectable filter, stability indicator and auto zero tracking
Analytical Balance,
Sartorius Analytical Balance
  • Capacity up to 320g
  • Readability up to 0.0001g (0.1mg)
  • Repeatability up to ±0.0001g (0.1mg)
  • Prices from £1230
  • Automatic calibration and adjustment with external weight
  • Electronic filters for adapting to ambient conditions
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007