Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Metering Pumps > Piston Pumps 
Pistons Pumps For Controlled Dispensing
  More Pumping Technology Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
High-Pressure Programmable Piston Pumps
• Drive features six dispensing modes
• Control speed, start/stop flow, or change fluid direction manually or remotely via analogue signal or RS-232 interface
piston pump
• Flow rates from 0.04 ml/min to 1300 ml/min
from  $3795.00   More details
Valveless Piston Pump Heads
• Pump fluids with viscosities up to 500 cp
• Flow range available for 0 to 1296 ml/min
valveless piston pump head
• Low dead volume
from  $435.00   More details
High Precision, High Pressure Piston Pumps
• Ideal for use in HPLC applications
• Pulse dampener significantly reduces pulsation with dead volume
high pressure piston pumps
• Pressure ratings up to 3000 PSI available
from  $1995.00   More details
Cole-Parmer UK offers a range of piston pumps suitable for controlled dispensing and metering applications. Piston pumps are highly accurate over long periods of operation and provide excellent repeatability. The piston pump can be run for long periods between maintenance making them suitable for continuous duty applications. Cole-Parmer’s piston pumps can deliver 0.04 mL/min to 1.3 LPM and deliver pressures up to 345 bar.
Cole-Parmer’s laboratory piston pumps are ideal for bench top use and include high pressure models for HPLC pumping applications as well as offering a wide range of control interfaces including analogue and RS-232 for connecting to a PC.
Our OEM and Industrial range of piston pumps use a valveless design. This creates a piston pump that offers enhanced reliability, simplifies troubleshooting, service and down time as there are no valves to clog and the moving parts in contact with the media is limited to just the piston. These piston pumps can deliver millions of cycles of high accuracy and drift free operation without the need of servicing.
Use Cole-Parmer’s on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Advantages for valveless piston pumps
  • Easy to use and virtually maintenance free
  • Excellent repeatability
  • Wide dynamic range
  • Viscosity independent
  • Choice of inert wetted parts including ceramic, 316 stainless steel and Kynar
  • Many control options including analogue, RS-232 and TTL inputs and outputs
Typical Application for piston pumps
  • Dispensing Systems – dispensing of solvents, UV adhesives, lubricants, reagents and mercury in the manufacture of electronics
  • Electronic Manufacturing – dispensing of electrolytes and slurries for batteries, dispensing of ceramic slurries in the manufacture of capacitors and diodes and the addition of flux for wave soldering equipment.
  • Environmental and Pollution Control – sampling stack gases, injection of monomers, polymers, and chemicals for water and waste treatment.
  • Food – for candy coating and polishing, vitamin fortification, addition of flavours, colours in food products
  • Fuel Cells – critical fluid control functions of the fuel cell operation including precision water addition, accurate water recirculation and liquid fuel metering.
  • Industrial Metering – including mixing of paint and pigment additives, catalyst for resins, plating bath regeneration, petroleum additives, photo chemicals, inks, monomers and adhesives.
  • Spraying Systems – including injection of insecticides, herbicides, and agricultural nutrients, as well as for ULV spray equipment.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007