Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Metering Pumps > Diaphragm Pumps 
Diaphragm Pumps and Diaphragm Pump Technology
  More Pumping Technology Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
Diaphragm Metering Pump
• Rugged, chemically resistant casing with choice of wetted parts
• Select from manual, remote or digital control models
Diaphragm metering pump
• Flow ranges: Liquid 0.4 ml/min – 76 LPH
from  $536.00   More details
Air-Powered Double Diaphragm Pumps
• High Flow rates up to 170 LPM
• Air-powered motor sparking concerns associated with electrical pumps
AODD pump
• Safely handles abrasive fluids and slurries - ideal for high viscousity and shear sensitive materials
from  $668.00   More details
Automatic-Demand Diaphragm Pumps
• Three-Chamber design provides smooth, pulse-free flow
• Flexible diaphragm design allows pump to run dry without damage
diaphragm pump
• Flow rates from 6.8 LPM
from  $93.00   More details
Atex-Approved Stainless Steel Double-Diaphragm Pumps
• Prevents electro-static discharge that could ignite in a gaseous explosive environment
• Flow rates of 400 LPM
atex aod pumps
• Viscosities up to 11,000 cp
from  $2704.00   More details
Diaphragm pump technology uses the force of a flexible diaphragm to displace liquid with each stroke. Using this principle, Cole-Parmer’s range of diaphragm pumps are able to transfer liquids with a flow rate of 0.07 mL/min to 560 LPM and pressures up to 30 bar.
Cole-Parmer offer three types of diaphragm pump technology with varying characteristics:
Digital remote contr
Solenoid Diaphragm Pump
  • Flow rates – 0.08 mL/min to 250 LPH
  • Pressures up to 17.3 bar
  • Use for controlled metering and dosing applications
Solenoid diaphragm pumps are known as metering pumps as they provide precise control of flow rates delivering from 0.08 ml/min up to 4,167 ml/min with pressures up to 17.3 bar. Our range includes manual and digital adjustable stroke lengths and stroke frequency to enable flow control. Pump models are available with external triggers and input control options and come in a variety of materials for the internals parts as well as the housing. These pumps requires no seals so you can pump abrasives and slurries if the valves are kept clean
Moderate Flow Positi
Positive Displacement Diaphragm Pump
  • Flow rates to 64 LPM
  • Pressures up to 30 bar
  • Use for general transfer and spraying applications
These types of diaphragm pumps are known as positive displacement pumps as they deliver a definite volume of liquid using one or two flexible diaphragms for each cycle of pump operation. Ideal for general transfer and spraying applications, our range can offer flow rates of up to 64 LPM, pressures up to 30 bar and with material options to allow compatibility with corrosive liquids. Many of our diaphragm pumps include factory set pressure limits to either shut the pump off or open a by-pass circuit within the pump to protect the pump and system from damage.
High-flow, air-drive
Air Operated Diaphragm Pump
  • Flow rates to 560 LPM
  • Pressures up to 8.3 bar
  • Use for general transfer applications and are ideal for transferring viscous and slurry media.
Air operated diaphragm pumps are often referred to as a AOD pumps or AODD pumps. They use the same technology as positive displacement diaphragm pumps however they contain no heat generating electrical components allowing safe, explosion-proof operation in hazardous areas. Used extensively in the food and paints industry for general transfer applications they are able to deliver flow rates up to 560 LPM. AOD pumps have the ability to handle low to high viscosity fluids and even particulates. They have the ability to self prime and are available in a variety of materials including PTFE and stainless steel.
Use Cole-Parmer’s on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007