Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
1-800-323-4340 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Metering Pumps > Syringe Pumps 
Syringe Pumps For Low Flow Rates
  More Pumping Technology Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Centrifugal Centrifugal pumps Shop online
Single Syringe Infusion Pump
• Microprocessor motor control and precision gearing
• Obtain precise flow rates down to 0.2µL/hour to 500ml/hour
syringe pump
• Accepts syringes from 10µL to 60ml
from  $1320.00   More details
Programmable Multi-Syringe Pumps
• Program flow rate hold, pause and chnage of directions
• Ten Syringes Pump
multichannel pump
• Flow rate per syringe 0.001µL/hr to 147ml/min
from  $3655.00   More details
Nano Syringe Pump
• Accepts syringes from 0.5µL to 250µL
• Remote injector and rugged mounting arm
nano syringe pump
• Preset rate and volume control
from  $2345.00   More details
Cole-Parmer UK’s syringe pumps are ideal to handle very low flow rates, ranging from 1nL/min to 147 ml/min. They move precise volumes of liquids in a specified time period to provide accurate flow rates.
Syringe pumps can operate is either infusion or withdraw mode to offer pulse free flow for a variety of applications. Syringes are available in plastic, stainless steel and glass to allow compatibility for applications including aggressive fluid transfer.
Cole-Parmer’s range of syringe pumps include programmable models to allow customised infusion and withdrawal patterns including commands to dispense a specified amount, automatic rate changes, programmed pauses, user or external inputs and sending and receiving signals from other equipment. Communication interfaces available include RS-232 and TTL signals. Our syringe pump technology includes models that can hold up to ten syringes, produce a force of up to 45kg and also includes economical OEM style pumps.
Use Cole-Parmer’s on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Advantages for syringe pumps
  • Easy to use and virtually maintenance free
  • Ideal for low flow at high pressure
  • Excellent pulseless flow
  • Available for OEM application
  • Available with accuracy of ±0.5%
  • Repeatability of ±0.2%
  • Wide dynamic range, typically 100,000:1
Typical Application for syringe pumps
  • Microfluidics/Nanofluidics
  • Neuroscience
  • Micro Dialysis
  • Drug/Nutrition Infusion
  • Cell Injection
  • Chemical Reactor Injection
  • Low flow dispensing
  • Aerosol Spray
  • Perfusion Chamber
  • Compound Storage
  • Secondary Screening
  • Primary Screening
  • Assay Development
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007