Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 30 pages on various tubing formulation along with hundreds of pages on fittings, flowmeters and other fluid handling products Request yours today.
Featured Products
Silicone (peroxide-cured) tubing - 1/8'' ID x 1/4''OD - 25 ft/pack
Silicone tubing
  • Sterilize: With autoclave or gamma irradiation
  • Temperature range: -60 to 460°F (-51 to 238°C)
  • Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic. Translucent
from $53.00 More details
PharMed BPT tubing - 1/4'' ID x 1/2'' OD - 500ft/pack
Pharmed Tubing
  • Ideal for applications including cell and tissue culture work
  • Temperature range: -75 to 275°F (-59 to 135°C)
  • Characteristics: nontoxic, opaque beige, extremely low permeability
from $1081.00 More details
PTFE tubing - 1/32'' ID x 1/16'' OD - 25 ft/pack
PTFE tubing
  • Sterilize: With autoclave, ethylene oxide, or use dry heat
  • Temperature range: -400 to 500°F (-240 to 260°C)
  • Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent
from $14.50 More details
Transfer Tubing
Cole-Parmer offer an extensive range of transfer tubing. Our flexible and rigid tubing formulations cover a wide range of characteristics to fulfil all your transfer tubing needs. Whether it is for everyday laboratory tubing applications or for industrial transfer applications we offer:
  • Tubing Size with inside diameters as small as 0.002 in (0.05 mm) to 2 in(50.8mm)
  • Pressure ratings up to 30 bar
  • Tubing to withstand Vacuum level of 759mm (29.9 in) of mercury at 23°C
  • Temperature ratings up to 288°C
  • Chemical compatible transfer tubing for the food, drink and pharmaceutical industry with our range of FDA approved material as well as tubing for aggressive chemicals and solvents.
  • Tubing suitable for sterilization techniques such as gamma irradiation, autoclaving and ethylene oxide sterilization
  • Tubing Resin Certification which available free of charge at time of order for a wide selection of our tubing. The certification states the tubing orders complies with regulatory standards.
  • Quantities for general use as well as spools of up to 150 m (500ft) for some our more popular ranges for production lines.
When selecting transfer tubing for your application, use our online technical tools or contact our application specialist to help you find the right tubing. We have over 50 years of experience and are always testing new liquids and gases for improved chemical compatibility ratings.
Cole-Parmer's Popular Transfer Tubing:
Silicone tubing
Peroxide-Cured Silicone Tubing
  • Flexible, soft and translucent tubing formulations
  • Available in platinum, peroxides cured and reinforced tubing formulations
  • Able to withstand temperatures up to 232°C
  • Sizing ranging from as small as 0.64 mm ID to 38.2 mm ID
  • Excellent biocompatibility with no leachable additives
  • FDA compliant making it ideal for pharmaceutical, biotech and food application
  • Available is standard pack sizes or spooled rolls for production runs
Tygon Tubing
Tygon lab tubing
  • Available in a range of formulations including Tygon Chemical, Tygon UV-Resistant, Tygon Vacuum and Tygon Fuel & Lubricant
  • Flexible tubing available in sizing from 0.19 mm to 38.1 mm
  • Temperature rating of up to 74°C
  • Long tubing life formulations available
  • Applications include general laboratory use, gasoline, hydrocarbons and alcohols
  • Available is standard pack sizes or spooled rolls for production runs
Viton Tubing
Masterflex Viton tubing
  • Ideal for high temperature work up to 204°C
  • Available in opaque white or opaque black
  • FDA approved tubing
  • Sizes from 0.89 mm ID to 19.1 mm
  • Ideal for transferring solvents and acids
  • Good chemical resistance
PTFE tubing
PTFE Tubing
  • Chemically inert, non-toxic and virtually nonporous
  • Temperature range of -240°C to 260°C
  • Ultra microbore tubing available 0.05 mm to 19 mm
  • FDA compliant tubing
  • Ideal for demanding lab, chemical, electrical and automotive applications
  • Translucent
  • Available is standard pack sizes or spooled rolls for production runs
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish