Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Featured Products
Masterflex Peristaltic Pumps
Peristaltic pump system
  • Complete dispensing systems with Tygon LFL tubing - up and running within minutes!
  • Flow rate 28 to 1,700 ml/min
  • Includes remote control options
from $1402.00 More details
Ismatec High-Accuracy Low Flow Pumps
ismatec pump
  • Up to 24 high accuracy channels in one pump
  • Analogue input and output controls
  • Flow rate per channel - 0.0012 to 8.86ml/min
from $3553.00 More details
Masterflex C/L Varible - Speed Tubing Pumps
CL pumps
  • Flow range: 0.001 to 39.4 ml/min depending on rpm and tubing size
  • Remove control connections on back of pump; start/stop pump with contact closure
  • Built-in tubing retainers firmly grip the tubing - change tubing quickly and easily
from $598.00 More details
Masterflex I/P Digital Brushless Process Drive
IP drive
  • Dispense by volume or time or pump continuously
  • Flow range: 0.01 to 26 LPM depending on rpm and tubing size
  • Brushless motor virtually eliminates maintenance
from $5145.00 More details
Tubing Pumps
Cole-Parmer offers a range of tubing pumps suitable for dispensing, metering and general transfer applications. Our tubing pumps offer flow rates as low as 0.0007 mL/min to 45 LPM and are able to generate pressures up to 8.6 bar (125 psi).
Tubing pumps confine the media to the tubing providing you with a pump that cannot contaminate the fluid and the fluid cannot contaminate your pump. This contamination free pumping makes tubing pumps great for use in high purity application including the transfer or dosing of chemicals and additives in food, pharmaceutical and semiconductor applications. In addition as the media is confined to the tubing a tubing pump offers easy maintenance and reduced down time compared to other pumping technologies.
Tubing pumps use flexible tubing to run through rollers in the pump head. As fluid goes through the pump head, packets of fluids are formed. Flow rate is determined by the size of the packets and the speed of the rollers turning. By controlling the size of the tubing and the speed of the pump head accurate metering of the media can be achieved.
Cole-Parmer offer tubing pumps suitable for use in labs, pilot plants, process and OEM applications. From low cost fixed speed pumps to advanced controlled models suitable for critical metering and dispensing applications Cole-Parmer is able to offer a tubing pump solution.
Browse through our on-line catalogue and use our tools to find your tubing pump or contact our application specialist for free technical support and let us bring over 50 years of pumping experience to solve your application needs.
Advantages for tubing pumps
  • The only part of a tubing pump that comes in contact with the liquid is the tubing. This maintains the sterility of fluid and the pump
  • Cole-Parmer offers over 20 tubing formulations and sizes for their tubing pumps for excellent compatibility with a variety of liquids
  • Self priming - our tubing pump will draw fluid into the tubing when starting dry, up to 8.8 m suction lift. Other pumps require user to fill pump and suction line with fluid before use which can be inconvenient and also create potential for hazardous spills or contamination
  • Gentle pumping action - low shear - makes a tubing pump ideal for handling cells and large proteins, as the pumping action will not damage the content
  • The fluid does not contact the tubing pump so there are no parts to clean such as valves, diaphragms, seals etc. All you have to do is clean or replace the tubing. This easy tubing replacement means reduced maintenance times
  • A tubing pump is non-siphoning which means that they prevent back flow into the system. This promotes accuracy during dispensing
Typical Application for tubing pumps
  • Juice production
  • Pizza sauce dispensing
  • Vitamin A & D Injection
  • Aspiration of tissue culture medium
  • Circulation of cell suspension in fermentation
  • Cosmetic dispensing
  • Harvesting cell media
  • Manufacturing IV bag dispensing
  • Nutrient supply for cultures
  • Spray coating dispensing
  • Sterilized media dispensing
  • Ultrafiltration
  • Acid/base dispensing
  • Adhesives for cement
  • Circuit board manufacturing
  • Dispensing glue emulsions
  • Transfer of Fuels and lubricants
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish