Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Featured Products
Siphon/Lift Pail Pump
hand operated pump
  • Suitable for pumping petroleum products or water
  • Cut inlet tube to fit your container
  • Maximum suction lift is 10ft
from $34.00 More details
Electric Drum Motor
Drum motor
  • Build a pump best suitable for your application - starting with this 9000 rpm motor
  • Use with our tube sets that can handle viscosities to 50,000 cp
  • Corrosion resistant expoxy coating resists caustic fluids
from $750.00 More details
Mixing Drum Pumps
pump
  • Air operated drum pumps
  • Mix and pump emulsions and suspensions in a single operation
  • Small pump tube diameter is ideal for use in applications with narrow necked containers
from $658.00 More details
Cole-Parmer Catalogue > Pumps > Hand and Drum Pumps
Hand Pumps and Drum Pumps
Cole-Parmer offers an extensive range in economical hand-operated pumps as well as electric driven and air-operated drum pumps. They are ideal for transferring fluids from drums or other containers up to 200 litres in capacity. Our range of hand and drum pumps are able to deliver flow rates of up to 238 LPM and pressures of 6.8 bar. They are ideal for delivering pulseless flow and are able to pump more viscous fluids up to 100,000 cp.
Hand pumps - Cole-Parmer offers several designs in hand operated pumps including siphon, piston, diaphragm and rotary hand-operated drum pumps. They are available in a range of materials from low cost plastics to stainless steel and PTFE, making them ideal for transferring non-corrosive chemicals such as oils, waxes and soaps as well as corrosives, detergents and acids.
Drum pumps - Cole-Parmer offers air operated as well as electric driven drum pumps. Our drum pump systems have a range of varying characteristics from short to long tube lengths for small to large containers, tube materials including plastic, PVDF, sanitary and stainless steel, motors that are explosion proof or air operated and batch controlled drum pumps to allow automatic dispensing of preset volumes.
Use Cole-Parmer's on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Advantages for hand and drum pumps
  • Different tube set materials to best suit your needs and applications
  • Adapters available for a range of containers
  • Options include variable speed motors, air operated and explosion proof motors
Typical Application for hand and drum pumps
  • Acid & Alkali drum pumps
  • Sanitary maintenance drum pumps
  • Petroleum drum pumps
  • Water & Utility pumps
Cole-Parmer Instrument Company Ltd.
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams
ABOUT SSL CERTIFICATES
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
View Popular Product Categories: mass flowmeters | tissue culture | vacuum pumps | pressure connectors | radiation detectors | pipette tips
View Top Selling Categories: peristaltic pump | laboratory equipment | chemical cabinets | test tubes | microbore tubing | ultrasonic cleaners
View Featured Categories: liquid nitrogen dewar | centrifugal pumps | centrifuge tubes | temperature humidity meter | petri dish