Home  >    

Thermo Scientific ERTCO Hydrometer, 9 to 21° Baume, 0.1° Divisions, Plain Form

WZ-08293-10
$34.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
12 10% $30.60/EACH
Discontinued
Qty
    Hydometers for use with liquids heavier than water.
    Manufacturer number 15059867
    Form Plain
    Baume range (min) (degrees) 9
    Baume range (max) (degrees) 21
    Length (mm) 300
    Model 2576
    Product Type Hydrometers, Degrees Baume
    Brand Thermo Scientific Ertco

    Accessories
    Qty/Price
    Availability

    at $51.00 USD/EACH

    In stock.
    Calibrations
    Qty/Price
    Availability

    at $135.00 USD/EACH
    Usually ships in 8 days.




    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought