Tag, Danger Out Of Service, Back A, 5 7/8" X 3 1/8", PF-Cardstock

WZ-40503-68
$18.00 USD/PACKAGE

Are you a reseller or buying in bulk? Call us today.


Usually ships in 11 days.
Qty
    OSHA/ANSI-Compliant Header Tags meet OSHA 29 CFR 1910.145 (f) and ANSI Z535.5 standards which provide specifications, designs, and guidelines for the use and application of accident prevention tags. These tags clearly indicate a temporary hazard that may lead to personal injury or property damage; use until that hazard is eliminated or hazardous operation is complete.

    All tags can be written on and have top gabled corners with 3⁄8" reinforced metal grommet on vinyl plastic.
    Product Type Accident Prevention
    Description OSHA/ANSI Tag
    Material Cardstock
    Legend Back side: DO NOT REMOVE THIS TAG! TO DO SO WITHOUT AUTHORITY WILL MEAN DISCIPLINARY ACTION! IT IS HERE FOR A PURPOSE
    Header DANGER
    Quantity 25
    Brand Accuform
    Model MDT158CTP
    Manufacturer number MDT158CTP





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title