Stir Bar Retriever, Rigid Polyethylene; 17" L
WZ-08555-40
$19.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
    Retrieve stir bars without messy spills or breakage. Alnico V magnet is completely enclosed in PTFE.
    Product Type Stir bars
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 International Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Do you have questions about this product?
    get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      Can these sticks be Autoclaved.
      Asked on 2/24/2010 by Anonymous from Centennial Co.
      1 answer
      • STAFF EXPERT
        A:
        These are made of polyethylene, and PE is not autoclavable.
        Answered on 2/25/2010 by Jen from Technical Support
    Displaying question 1
    Do you have a question about this product? 
    Product Reviews
    REVIEW SNAPSHOT®
    by PowerReviews
    Stir Bar Retriever, Rigid, 17"L
    5.0
    (based on 1 review)
    Reviewed by 1 customer
    Displaying review 1
    Back to top
    5.0
    I WOULD BUY THIS PRODUCT AGAIN
    By NUVIS
    from SOMERSET,NJ
    About Me Lab Assistant
    Verified Buyer
    Pros
    • Easy To Use
    • Works Properly
    Cons
    • None
    Best Uses
    • In The Lab
    • Research
    Comments about Stir Bar Retriever, Rigid, 17"L:
    WE USE THIS PRODUCT IN THE LAB.
    WORKS GREAT
    Displaying review 1
    Back to top
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title