Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

STACO L501B: Fully enclosed variable-voltage controller; 120 VAC in, 0 to 140 V out, 4.5 A

WZ-01575-22
$502.00 USD/EACH

Usually ships in 9 days.
Qty

  • Minimize voltage variations from no-load to full-load current conditions
Reduce the need to readjust the output voltage as load changes. Simply turn the control knob clockwise to increase output linearly. Unlike solid-state controls, these units do not cause waveform distortion—eliminating errors in voltage and current measurement.
Select a ventilated, steel cased model for your benchtop applications or general utility mounting.
Fully-enclosed models offer maximum safety and protection from physical abuse, chemical spills, and other hazards. Flat-top enclosures with recessed control panel protects against chemical spills. Mount units on the wall, inside other equipment, or use as portable units.
What's included: a 6-ft grounded cord with plug, a three-conductor receptacle, pilot light, fuse, and a double pole on/off switch which disconnects both sides of the line to prevent electrical shock.
Product Type Variable Voltage Controllers
Max amps 4.5
Input (VAC) 120
Input (Hz) 50/60
Output (VAC) 0 to 140
Brand Staco





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title