Pressure Transducer, I to P, 4-20MA-3-120PSI I/P (Curr To Press)
WZ-68458-24
$372.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 15 days.
Qty
  • High-flow capacity up to 12 scfm
  • 4 to 20 mA Input
  • Mounts in any angle
  • Small size and lightweight
  • External zero and span adjustments
These transducers are an electro-pneumatic device that reduces supply pressure to a regulated output pressure directly proportional to an electrical input signal. These transducers are also field-reversible by reversing the polarity of the signal leads and recalibrating. After calibration, the transducer will then provide a minimum of it''s full rated output pressure upon input signal failure. These units are typically used in HVAC-R systems, petrochemical processing, and slurry processing.
Product Type I/P transducers
Brand Marsh Bellofram
CE Compliance Yes
Manufacturer number 961-111-000
Model 961-111-000
Accessories
Qty/Price
Availability
at $26.50 USD/EACH
Usually ships in 15 days.
at $77.00 USD/EACH
Usually ships in 15 days.
at $143.00 USD/EACH
Usually ships in 15 days.
at $204.00 USD/EACH
Usually ships in 15 days.
at $9.00 USD/EACH
Usually ships in 15 days.
Calibrations
Qty/Price
Availability
at $155.00 USD/EACH
Usually ships in 6 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title