Merck Index, 14th Edition

WZ-00561-32
$193.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
10 10% $173.70/EACH
25 15% $164.05/EACH
50 20% $154.40/EACH
    This updated encyclopedia of chemicals, drugs, and biologicals includes a therapeutic category, biological activity index to drugs, CAS Registry Number, substance indices, and more. Also available on CD-ROM for WINDOWS ®  or MACINTOSH ®  with quick access to databases via multiple search options.



    Product Type Biochemistry Books
    Author Edited by Maryadele J. O'Neil
    No. of pages 2564
    ISBN 978-0-911910-00-1
    Edition 14th Edition
    Published date 2006





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    Merck Index, 14th Edition
     
    5.0

    (based on 1 review)

    Reviewed by 1 customer

    Displaying review 1

    Back to top

    (1 of 1 customers found this review helpful)

     
    5.0

    Great Reference

    By Hazmatstorm

    from Rogers, Arkansas

    About Me Student

    Verified Buyer

    Pros

    • Accurate

    Cons

    • None

    Best Uses

    • In The Field
    • In The Lab
    • Research

    Comments about Merck Index, 14th Edition:

    A great reference. anyone involved in the use of chemicals should have this close by!

    Displaying review 1

    Back to top

    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title