Kimberly-Clark KLEENGUARD V30 Safety Glasses, Anti-Fog Clear Lens

WZ-15940-61
Not available in USA & Canada.

  • Polycarbonate lens provide a light-weight, comfortable fit
  • Available in a variety of options - V10, V20, V30 and V40 safety glasses
  • Impact resistant
  • Offer 99.9% UVA/UVB protection
KLEENGUARD safety glasses are impact resistant and provide 99.9% UVA/UVB protection.
All glasses meet ANSI Z87.1+ and EN 166 standards.
KLEENGUARD V10 Glasses feature lightweight frames and a choice of clear, amber, or clear OTG (over the glasses) lens.
KLEENGUARD V20 Glasses have wraparound lenses for better eye coverage and padded temples for added comfort. Choose from smoke, anti-fog clear, and indoor/outdoor lenses.
KLEENGUARD V30 Glasses feature wraparound lenses along with padded nose bridge and temples and adjustable lens angle. Choose from amber, anti-fog clear, and indoor/outdoor lenses.
KLEENGUARD V40 Glasses are the ultimate in comfort. Dual-molded rubber temples and cushioned brow bar give a comfortable feel while adjustable soft nose bridge and lens angle allow you to customize the fit. Silver mirror lens is perfect for outdoor environments.
Product Type Glasses
Color Clear
Lens Clear/Anti-fog
Compliance ANSI Z87.1, EN 166
Model 08452
Brand KLEENGUARD
Manufacturer number 08452





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title