Traceable Monitoring Hygrometer
Representative image only.
Traceable Monitoring Hygrometer
Traceable | Mfr: 6040TR| Item: 2025030
Simultaneously displays time-of-day, humidity and temperature
£50.44GBP / Each
£60.53GBP (incl VAT)
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
£50.44GBP / Each
£60.53GBP (incl VAT)
Buy It Together
Current Item
Traceable Monitoring Hygrometer
Traceable Monitoring Hygrometer
2025030
£50.44GBP / Each
£60.53GBP (incl VAT)
+
Traceable Ultra™ Refrigerator/Freezer Thermometer; 1 Bottle Probe
9876754
£74.42GBP / Each
£89.30GBP (incl VAT)
+
Traceable Jumbo Fridge/Freezer Digital Thermometer; 1 Bottle Probe
9446082
£80.94GBP / Each
£97.13GBP (incl VAT)
+
Traceable Humidity Card with Calibration; 6/PK
0331306
£69.03GBP / Pkg of 6
£82.84GBP (incl VAT)
+
Traceable Fridge/Freezer Digital Thermometer; 1 Bottle Probe
9446072
£79.92GBP / Each
£95.90GBP (incl VAT)
+
Traceable Excursion-Trac® Data Logging Thermohygrometer/Barometer
9876715
£272.48GBP / Each
£326.98GBP (incl VAT)
+
Traceable Three Channel Alarm Timer
9441110
£64.60GBP / Each
£77.52GBP (incl VAT)
+
Traceable Big-Digit Memory Thermometer, 1 Bullet Probe
9000075
£59.81GBP / Each
£71.77GBP (incl VAT)
+
Traceable Memory-Loc™ Data Logging Thermohygrometer
9876716
£276.63GBP / Each
£331.96GBP (incl VAT)
+
Digi-Sense Thermocouple Connector, Mini, Type-T, Female Jack; 5/pk
1852700
£31.54GBP / Pkg of 5
£37.85GBP (incl VAT)
=
Total Price 1059.81 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Traceable® Certificate, built-in stand, wall mount, and battery.
Key Features
  • Recall the highest and lowest temperature and humidity readings at the touch of a button
  • Display time in 12-hour or 24-hour military time format
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
This calibrated hygrometer provides precision environmental monitoring in laboratories, cleanrooms, warehouses, medical storage facilities, and food production environments. Designed for durability and ease of use, the hygrometer features a large digital display showing the temperature, humidity, and time, making it easy to view the measurements at a glance. Its compact, battery-operated design allows for flexible placement, whether wall-mounted or used as a portable device.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
Conformite EuropeenneNIST Traceable Calibration1 Year Warranty
Specifications
  • Min RH Range (%)20
  • Max RH Range (%)90
  • RH Accuracy±5% from 40 to 70% RH; ±8% otherwise
  • RH Resolution (%)1%
  • Min Temperature (° F)32
  • Min Temperature (° C)0
  • Max Temperature (° F)122
  • Max Temperature (° C)50
  • Temperature Accuracy±1°C from 0 to 40°C
  • Temperature Resolution0.1°
  • CertificationsNIST-Traceable
  • Battery1 AAA (included)
  • Height (in)4 9/16
  • Width (in)2 3/4
  • Depth (in)9/16
  • DescriptionNIST Calibrated Monitoring Hygrometer