Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

CoorsTek High-Alumina Rectangular Tray, 750 mL, 188 x 140 x 42 mm; 1/Pk

CoorsTek  |   Mfr# 65582   |   Item# WZ-17826-12

$297.00 USD/CASE

Quantity Discounts
Qty: 5 10
Price: $267.30 $252.45
    Trays are 99.8% pure aluminum oxide. Highly resistant to the effects of chemicals, these trays are ideal for chemists, metallurgists, and others who are involved in high-temperature work and need contamination-free results. Autoclavable.
    Product Type Trays
    Material Aluminum Oxide
    Capacity (Liters) 0.75
    Dimensions 188 mm L x 140 mm W x 42 mm H
    Qty/ea 1
    Model 65582
    Brand CoorsTek
    Manufacturer number 65582

    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - International
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:

      Do you offer a lid for the tray?
      Asked on 3/23/2011 by Anonymous

      1 answer



        No, there is currently not a lid for this tray. We have some other chemically resistant containers that are offered with lids. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).

        Answered on 3/25/2011 by Jack from Technical Support
    • Q:

      What is the highest temperature these can be used? Also, what is the highest ramp rate they can handle?
      Asked on 3/2/2011 by Anonymous

      1 answer

      • Staff Reviewer


        This tray can tolerate up to 1750 degrees Celsius however the rate of heating and cooling is important. These can only handle a temperature change of 150 degrees Celsius per hour.

        Answered on 3/4/2011 by Jen from Technical Support
    Displaying questions 1-2

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought