Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Compression Fitting, Reducing Union, 316 SS, 3/8" x 1/4" OD
Item# WZ-31406-25
$20.00 USD/EACH
Quantity Discounts
Qty: 10 50 100+
Price: $18.00 $17.00 $16.00
Usually ships in 9 days.
Qty
    Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
    Product Type Compression Fitting
    Material Stainless Steel
    Tube OD (in.) 0.375 - 0.25
    Sterilization Ethylene Oxide, autoclave, or gamma
    Qty/ea 1
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - International
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      Does this contain any lead?
      Asked on 2/25/2014 by Fred
      1 answer
      • STAFF EXPERT
        A:
        No, it is lead free
        Answered on 2/27/2014 by Anthony from Technical Support
    Displaying question 1
    Do you have a question about this product? 
    Product Reviews