Cole-Parmer Fermentation DO Probe; Glass/Polarographic/180 mm

WZ-05643-00
$219.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


  • Low-cost, glass-body design
  • Polarographic
These economical glass-body probes are ideal for your laboratory fermentation applications where sterilization takes place in a separate autoclave. Probes feature a threaded detachable cable lead. Order appropriate cable separately.
Probe type Polarographic
Maximum pressure 2.4 bar (35 psi)
Response time 98% in 60 seconds
Body material Glass
Connector threaded detacheable lead
Dimensions 180 x 12 mm
Brand Cole-Parmer

Accessories
Qty/Price
Availability

at $63.00 USD/EACH

Usually ships in 12 days.

at $63.00 USD/EACH

Available for order.
Need it sooner?

at $60.00 USD/EACH

Available for order.
Need it sooner?

at $14.50 USD/EACH

Available for order.
Need it sooner?

at $38.00 USD/EACH

Available for order.
Need it sooner?

at $37.00 USD/EACH

Available for order.
Need it sooner?

at $31.00 USD/EACH

Available for order.
Need it sooner?




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

10 Questions | 10 Answers
Displaying questions 1-10
  • Q:

    Please tell me what kind of output signal does it have.
    Asked on 3/20/2014 by vio

    1 answer

    • STAFF EXPERT

      A:

      It gives off an amp signal that usually goes to a Oxygen meter or controller.

      Answered on 3/21/2014 by Anthony from Technical Support
  • Q:

    For the membrane replacement, can I replace the methyl chloroform with other solvents? The methyl chloroform is so expensive and hard to buy. Thanks!
    Asked on 2/19/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      The only solvent recommended that has been verified to work is the methyl chloroform solvent. Other solvents may work but that would be up to you to determine and we would not be responsible for damage that may occur to the membrane. This would be at your own risk if you decide to test other solvents to soften the membrane.

      Answered on 2/20/2014 by Jose from Technical Support
  • Q:

    What does the "Membrane Service Kit" include? Does it include the methyl chloroform solvent mentioned in the instruction? Is the methyl chloroform solvent equal to 1,1,1-Trichloroethane? I am wondering whether the "1,1,1-Trichloroethane in DMSO" is OK for this membrane replacement process. Thanks!
    Asked on 2/12/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      This kit contains: 3x 19mm membranes, o-rings, and 125mL fill solution. It does not include methyl chloroform solvent that is recommended to soften the silicone tubing, which is also not included.

      Answered on 2/12/2014 by Jack from Technical Support
  • Q:

    What's the difference between 1-mil and 2-mil PTFE membranes (EW-05643-50 and EW-05643-51). How to decide which one should I buy? Thanks!
    Asked on 2/6/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      The 1mm is going to have a faster response, but will be a little more fragile than the 2mm version. 2mm membrane is good in a more aggressive fermentation application.

      Answered on 2/6/2014 by Anthony from Technical Support
  • Q:

    Does this probe need to refill the solution frequently? What kind of Membrane Service Kit do I need for this probe? 1-mil thick PTFE Membranes or 2-mil thick PTFE Membranes? Does this probe need to be refill or change membrane frequently? Thanks!
    Asked on 2/6/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      The frequency that fill solution will vary depending on use from application to application. You will need to check to make sure the fill solution is filled accordingly. The fill solution is available as 05663-54. Either the 1-mil and 2-mil PTFE membranes can be used.

      Answered on 2/6/2014 by Jose from Technical Support
  • Q:

    Please recommend a DO meter that compatible with this DO probe. Thanks!
    Asked on 1/30/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      Item number 00336TN, together with power cord 50001-68, make up the discontinued benchtop DO meter 01971-00 that is compatible wit this line of probes.

      Answered on 1/31/2014 by Jose from Technical Support
  • Q:

    How many times usually this probe can be autoclaved? Thanks!
    Asked on 1/29/2014 by John from Blacksburg, VA

    1 answer

    • STAFF EXPERT

      A:

      There is no specific data on the number of autoclave cycles that this is rated for. They are designed to withstand repeated autoclaving and the actual number of cycles will vary.

      Answered on 1/30/2014 by Jose from Technical Support
  • Q:

    is it possible to autoclave it?
    Asked on 10/28/2010 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      This is a steam sterilizable sensor. As long as the cap is placed on the connector end, the sensor can be autoclaved.

      Answered on 11/2/2010 by Jen from Technical Support
  • Q:

    Could you please let me know if this DO electrode can work with the DO200 Polarographic controllers, 53405-40 or 53405-42. Does the electrode compensates for temperature?

    Thanks
    Asked on 12/28/2009 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      This probe does not offer temperature compensation and we do not suggest its use with either the 53405-40 or -42.
      Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).

      Answered on 12/29/2009 by Jen from Technical Support
  • Q:

    What is the procedure to replace the membrane?
    Asked on 6/12/2009 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      Please contact our technical application specialists for additional assistance and documentation.

      Answered on 6/19/2009 by Jen from Staff Expert
Displaying questions 1-10

Do you have a question about this product? 

Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
Cole-ParmerPolarographic Probe, 180 mm Immersion Depth
 
1.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
1.0

Probe not working

By Dave

from Minneapolis,MN

About Me Student

Pros

    Cons

    • Did Not Work

    Best Uses

    • In The Lab
    • Research

    Comments about Cole-Parmer Polarographic Probe, 180 mm Immersion Depth:

    I received this product and it was defective. I was unable to get a signal from it using a BNC connection. I used an other version to make sure it was indeed the probe. I asked for an exchange for another probe but the new probe that was sent was also defective. The electrolyte solution was leaking from the probe itself.

    Displaying review 1

    Back to top