Cole-Parmer Electrophoretic Blotting System, 4 Cassette capacity

WZ-28580-20
$645.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


  • For Western, Southern, or Northern blots—transfer proteins and nucleic acids from agarose or acrylamide gels
Glass cooling chamber with tubing adapters is built into base for temperature-controlled runs. Platinum wire electrodes are mounted in recessed grooves for protection from damage when cleaning unit or loading cassettes. Electrode design provides even current distribution over entire gel surface. Place a magnetic stirring bar beneath cassettes to circulate buffer within unit for more efficient cooling. Order additional cassettes separately.
What's Included:
safety cover with attached power leads, two platinum wire electrode panels, one gel cassette assembly (four for model 28580-20), internal glass cooling base, and tubing adapters.
Buffer solution required (mL) 1100
Cell dimensions (" W) 5.13
Cell dimensions (" H) 6.3
Cell dimensions (" D) 5.5
CE Compliance Yes
Product Type Electrophoresis Vertical Systems
Brand Cole-Parmer
Manufacturer number EBU204

Accessories
Qty/Price
Availability

at $9.50 USD/EACH

In stock.

at $40.00 USD/EACH

Available for order.
Need it sooner?




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    What material is the tank made of?
    Asked on 8/25/2010 by Alex from KS

    1 answer

    • STAFF EXPERT

      A:

      The tank is made of Acrylic.

      Answered on 8/26/2010 by Mike from Tech Support
Displaying question 1

Do you have a question about this product? 

Product Reviews