Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Laboratory Equipment > Industrial Mixers  
Laboratory Stirrers
  More Information Buy
laboratory Laboratory Mixers Shop online
industrial Industrial Mixers Shop online
handheld Handheld Mixers Shop online
mixer Accessories for Industrial Mixers Shop online
laboratory Accessories for Laboratory Mixers Shop online
Compact Digital Lab Mixer
• Use virtually anywhere - compact design with panel light
• Stir up to 25 liters and a maximum visc ousity of 15,000 ps
digital lab mixer
• Four button control - power on/off, speed increase/decrease, light on/off
from  Call for price   More details
General Purpose LabMaster Mixers
• Available in 3 mounting styles
• Available in rpms up to 2300
laboraory mixer
• Choice of fixed or remote control speeds
from  Call for price   More details
Cimarec Stirrers
• Available in top plates sizes of 100 mm, 175 mm and 250 mm
• Speed ranges up to 1200 rpm
laboratory stirrers
• Stirring capacities up to 20L
from  Call for price   More details
All-in-one Shaker
• Rocking motion: 10 to 70 cycles/min
• Rotating motion: 5 to 35 rpm
laboratory shaker
• For use with tubes and bags with optional expansion kits
from  Call for price   More details
Cole-Parmer offers an extensive range of laboratory stirrers and mixers for mixing and blending applications in the laboratory. Our mixing technologies include overhead laboratory mixers, magnetic stirrers and laboratory shakers. When considering your laboratory mixing needs, consideration has to be given to a range of factors. View our online catalogue to see our full selection of products or contact our Application Specialist to help you find the right product for your needs.
Overhead Laboratory Mixers
An overhead mixer works by placing an impeller in to a vessel containing the media to be mixed or blended. The impeller is driven by a motor connected to the impeller via a shaft. Laboratory mixers are used to mix, emulsify, homogenize, disintegrate and dissolve samples. Cole-Parmer’s large selection of overhead laboratory mixers include fixed and variable speed options, with rotational speeds up to 10,000 rpm and a choice of control functions including reversing and timed cycles. For applications requiring profiling select our computer compatible lab mixer.
overhead mixers
Features available with the Cole-Parmer model range of laboratory mixers:
  • Fixed or variable speed
  • Digital or analogue control
  • Optional computer control functionality
  • Electric and air driven laboratory mixers
  • Range of mixer impellers for a variety of mixing applications
Laboratory Stirrers
Laboratory magnetic stirrers work by placing a stir bar (a small magnet normally coated in a plastic such as PTFE) in to the vessel containing the media to be stirred. The stir bar is then rotated by using a plate containing rotating magnets or stationary electromagnets. Cole-Parmer’s range of laboratory magnetic stirrers can be used with a range of vessels including beakers, flasks and test tubes.
laboratory stirrers
Features available with the Cole-Parmer model range of laboratory magnetic stirrers:
  • Fixed or variable speed ranges
  • Digital or analogue control
  • Single or multiple position stirrers
  • Stirring hot plates
  • Submersible models
  • Plate material including ceramic, aluminum, stainless steel and glass
  • Programmable digital models
Laboratory Shakers
Laboratory shakers are used to blend or agitate samples within a vessel. The shaker has an agitation platform connected to a motor and control panel. The platform is designed to take beaker, flasks or tubes and mix the sample via the shaking motion of the platform. Laboratory shakers are categorized by their manner of shaking motion and include orbital, reciprocating, rocking, rolling, rotating and vortex. These laboratory shakers are available with a range of features including temperature control; timer control; single and multi-vessel holders; fixed and variable speed models.
laboratory shakers
Features available with the Cole-Parmer model range of laboratory mixers:
  • Orbital/Rocking/Vortex/Rotating shaking actions
  • Digital or analogue models
  • Adapters available for a range of vessels
  • Temperature control models
  • Programmable digital models
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007