Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Flow Meters > Variable Area Flow Meters 
Variable Area Flow Meters
  More Information Buy
Variable Variable Area Shop online
gear Gear Flowmeters Shop online
Turbine Turbine Flowmeters Shop online
Magnetic Magnetic Flowmeters Shop online
Gas Gas Mass Flowmeters Shop online
Differential Differential Pressure Flowmeters Shop online
Ultrasonic Ultrasonic Flowmeters Shop online
Variable Area Flowmeters
• Low cost acrylic body
• Choice of direct reading meters, with or without control valve
va meter
• Air: 0.04 LPM - 3,400 LPM Water: 5 ml/min - 75 LPM
from  Call for price   More details
Multi-tube Systems For Mixing Applications
• Choice of direct reading meters, with or without control valve
• Create your own multi tube or gas proportioner
multi flowmeter
• Available for Air, Argon, Carbon Dioxide, Helium, Hydrogen, Oxygen and Water
from  Call for price   More details
Cole-Parmer offer an extensive range of variable area flow meters for both liquid and gas applications. Cole-Parmer’s variable area flow meters are simple to install and offer cost effective solutions for applications requiring visual indication of flow or flow rate. Cole-Parmer offers variable area flow meters for measuring liquids flow from 0.0002 ml/min to over 400 LPM and for gases from 0.02 ml/min to over 700 m3/hour.
Cole-Parmer’s variable area flow meters are available in a wide range of materials including low cost acrylic and specialist options such as PTFE for aggressive media or stainless steel for high temperature and pressure applications. Many of our variable area flowmeters come with a wide range of control and output options including control valves, set-point switches and analogue outputs.
To find the right flow meter for your application use Cole-Parmer’s on-line flow meter quick search. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs.
Key Points for variable area flow meters
  • Variable area flow meters operate on the principle that the flow of the media will alter the position of a float in a tapered tube. The position of the float in the tapered tube is proportional to the volumetric flow rate
  • Typical accuracy of variable area flow meters is ±3% full scale
  • A variable area flow meter normally has a turn down ratio in the region of 10:1
  • A variable area flow meter normally requires vertical installation and a straight pipe run (minimum length of five times the diameter of the pipe) before the flow meters inlet
Advantages for variable area flow meters
  • Low cost
  • Simple to install
  • Easy to use and virtually maintenance free
  • Wide choice of materials including all PTFE for aggressive fluids
  • Many control options including control valves, set-point switches and analogue outputs
Disadvantages for variable area flow meters
  • Lower accuracy compared to many other flow meter technologies
  • Affected by pulsation
  • Sensitive to fluid changes such as viscosity, density and temperature
Typical Application for variable area flow meters
  • Liquid flow meter or gas flow meter where high accuracy is not required
  • Measuring water and gas flow in plants or labs
  • Monitoring chemical lines
  • Purging instrument air lines (i.e. lines that use a valved meter)
  • Monitoring filtration loading
  • Monitoring flow in material-blending applications (i.e. lines that use a valved meter)
  • Monitor makeup water for food & beverage plants
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007