Product Info
Product Info        
Brand YSI
Item Model DO200 Kit
Product Type Dissolved Oxygen Meter
Unit Of Measure Each
Number of Items 1
Warranty Period Unit 1 Year
Product Name YSI EcoSense DO200A Dissolved Oxygen Meter
Technical Specs
Technical Specs        
Meter Type Portable
Display type LCD
Min Dissolved Oxygen 0 to 20 ppm; 0 to 200%
Dissolved Oxygen Resolution 0.01 ppm; 10% saturation
Dissolved Oxygen Accuracy ±2% FS
Min Temperature (° C) -5
Max Temperature (° C) 50
Min Temperature (° F) 23
Max Temperature (° F) 122
Waterproof Yes
Probe Included No
Warranty 1 year
Compliance
Compliance        
Agency Compliance Prop 65 NA (no Prop 65 chemicals)