Product Info
Product Info        
Brand IKA
Item Model Eurostar 60
Product Type Overhead Mixer
Unit Of Measure Each
Number of Items 1
Warranty Period Unit 2 Years
Product Name IKA EUROSTAR 60 Control Digital Mixer with Wireless Remote
Technical Specs
Technical Specs        
Min Speed (rpm) 30
Max Speed (rpm) 2000
Input Power (watts) 168
Through-Shaft Yes
Chuck Chuck
Max Torque (in-oz) 85
Remote Control Yes
Display Yes
Power (Phase) 1
Power (VAC) 115
Power (Hz) 50/60
Motor hp 3/16
Warranty 2 Years