Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456

More Unique Lab & Industrial Supplies

 

Syringe Pumps

Syringe Pumps

Cole-Parmer's syringe pumps are ideal to handle very low flow rates, ranging from 1.3pL/min to 147 ml/min. They move precise volumes of liquids in a specified time period to provide accurate flow rates.
Syringe pumps can operate is either infusion or withdraw mode to offer pulse free flow for a variety of applications. Select from plastic syringes, stainless steel syringes or glass syringes.

Cole-Parmer's range of syringe pumps include programmable models to allow customised infusion and withdrawal patterns including commands to dispense a specified amount, automatic rate changes, programmed pauses, user or external inputs and sending and receiving signals from other equipment. Communication interfaces available include RS-232 and TTL signals. Our syringe pump technology includes models that can hold up to ten syringes, produce a force of up to 45kg and also includes economical OEM style syringe pumps.

Use Cole-Parmer's on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.

Advantages for syringe pumps

  • Easy to use and virtually maintenance free
  • Ideal for low flow at high pressure
  • Excellent pulseless flow
  • Available for OEM application
  • Available with accuracy of 0.5%
  • Repeatability of 0.2%
  • Wide dynamic range, typically 100,000:1

Typical Application for syringe pumps

  • Microfluidics/Nanofluidics
  • Neuroscience
  • Micro Dialysis
  • Drug/Nutrition Infusion
  • Cell Injection
  • Chemical Reactor Injection
  • Low flow dispensing
  • Aerosol Spray
  • Perfusion Chamber
  • Compound Storage
  • Secondary Screening
  • Primary Screening
  • Assay Development
Need
Assistance?

Get help from
our Experts!
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price

Cole-Parmer single-syringe infusion pump, 230 VAC

  • Product Type: Syringe Pump
  • Reproducibility: ±0.2%
  • Accuracy: ±.5%
WZ-74900-05
In stock.
$1,576.00 USD/EACH

Qty:

Cole-Parmer Syringe Pump, Infusion Only, Touchscreen Control 74905-50

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 100 to 240 VAC
WZ-74905-50
In stock.
$3,026.00 USD/EACH
$2,600.00 USD/EACH

Qty:

Cole-Parmer dual-syringe infusion pump, 230 VAC

  • Product Type: Syringe Pump
  • Reproducibility: ±0.1%
  • Power: 230 VAC
WZ-74900-15
Usually ships in 10 days.
$2,839.00 USD/EACH

Qty:

Cole-Parmer Four Syringe Pump, Touchscreen Control, Infuse/Withdraw

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 110 to 240 VAC
WZ-74905-39
Usually ships in 14 days.
$4,282.00 USD/EACH

Qty:

Cole-Parmer Syringe Pump, Infusion Only, Touchscreen Control 74905-02

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 100 to 240 VAC
WZ-74905-02
In stock.
$1,800.00 USD/EACH

Qty:

Cole-Parmer Syringe Pump, Infusion and Withdrawal Programmable, Touchscreen Control

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 100 to 240 VAC
WZ-74905-54
In stock.
$3,988.00 USD/EACH
$3,589.20 USD/EACH

Qty:

Cole-Parmer Syringe Pump Lubricant Super Lube (1cc)

  • Product Type: Maintenance tool
  • Qty/ea: 1
  • Model: 788316
WZ-74905-86
Usually ships in 7 days.
$8.00 USD/EACH

Qty:

WZ-74901-91
Usually ships in 5 days.
$2,351.00 USD/EACH

Qty:

Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe Pump

  • Product Type: RS-232 Cable
  • Qty/ea: 1
  • Brand: Cole-Parmer
WZ-74901-88
Usually ships in 8 days.
$148.00 USD/EACH

Qty:

Cole-Parmer Four Syringe Picoliter Pump, touchscreen control

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 110 to 240 VAC
WZ-74905-19
Usually ships in 5 days.
$4,303.00 USD/EACH

Qty:

Cole-parmer Stainless Steel Syringe, 50 mL, Viton O-ring, 1/8

  • Material: Stainless Steel
  • Product Type: Syringe
  • Capacity: 50 mL
WZ-74044-32
Usually ships in 7 days.
$753.00 USD/EACH

Qty:

Cole-Parmer Syringe Pump, Infuse / Withdraw Programmable, Touchscreen Control

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 100 to 240 VAC
WZ-74905-04
In stock.
$2,347.00 USD/EACH

Qty:

WZ-74901-90
Usually ships in 10 days.
$103.00 USD/EACH

Qty:

Cole-parmer Stainless Steel Syringe, 8 mL, Viton O-ring, 1/16

  • Material: Stainless Steel
  • Product Type: Syringe
  • Capacity: 5 mL
WZ-74044-10
Usually ships in 14 days.
$868.00 USD/EACH

Qty:

Cole-Parmer Touchscreen Syringe Pump; 2-Syringe/Infuse/Withdraw

  • Product Type: Syringe Pump
  • Reproducibility: ±0.05%
  • Power: 100 to 240 VAC
WZ-74905-08
Usually ships in 12 days.
$2,375.00 USD/EACH

Qty:

91 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#