Subtotal = £0.00 
Items = 0 
View Cart
United Kingdom
0500 345 300 or 
Welcome,
log in or register
You are filtering by:
Category:
Clear Category Laboratory Supplies & Consumables
Clear Category Spatulas & Stir Rods
Clear all filters
Category:
Brand:
Product Type:
Price Range:
Availability:
Sterile:
Yes (30)
Material:
Glass (11)
Capacity (mL):
0 - 20 (3)
Readability (g):
0.1 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 0500 345 300
Home  >  Laboratory Supplies & Consumables > Spatulas & Stir Rods
Spatulas & Stir Rods
Spatulas & Stir Rods
A lab spatula has many uses including: scooping and spreading. Micro is a lab spatula that is excellent for handling extremely small samples. A PTFE coated lab spatula is ideal for handling chemicals. A nickel or stainless steel spatula is versatile and when coated with PP they feature excellent chemical and heat resistance. A lab spatula may be single use, sterile and non-sterile. There are also a myriad of stirring rods. These vary in length, tip shape and materials. The ones coated with PP and PTFE are autoclavable and provide excellent heat resistance 266°F (130°C for PP and 550°F (287°C) for PTFR. PE/Steel offers excellent chemical resistance and temp resistance -58 to 176°F (-50 to 80°C). Glass rods use soft borosilicate glass, offer chemical resistance and can withstand temperatures to 400°F (205°C). Many have policemen to help transfer liquids or clear out precipitate. If you have questions about selecting the correct lab spatula or stirring rod for your application speak with one of our Technical Experts.
 Product Detail
Sort
 Availability
 Price
Representative photo only SMART SPATULA Disposable Spatula blue 210 mm
SMART SPATULA Disposable Spatula, blue 210 mm
Product Type Spatulas,Material Polypropylene,Brand Smart Spatula
WZ-06265-18
In stock.
£26.80 GBP/PACKAGE
£32.16 (incl VAT)
Qty:
WZ-06369-16 Nickel/stainless steel laboratory spatula, with 1 1/8"L spooned end and 1 5/8"L flat squared end
Nickel/stainless steel laboratory spatula, with 1 1/8"L spooned end and 1 5/8"L flat squared end
Product Type Spatulas,Material Nickel/Stainless Steel,Sterile Yes
WZ-06369-16
In stock.
£24.56 GBP/PACKAGE of 3
£29.47 (incl VAT)
Qty:
Representative photo only SMART SPATULA Disposable Spatula Opaque 210 mm
SMART SPATULA Disposable Spatula Opaque 210 mm
Product Type Spatulas,Material Polypropylene,Brand Smart Spatula
WZ-06265-12
Usually ships in 7 days or sooner.
£25.46 GBP/PACKAGE
£30.55 (incl VAT)
Qty:
Representative photo only SMART SPATULA Disposable Macro Spatula Green 310 mm
SMART SPATULA Disposable Macro Spatula, Green 310 mm
Product Type Spatulas,Material Polypropylene,Brand Smart Spatula
WZ-06265-16
In stock.
£25.68 GBP/PACKAGE
£30.82 (incl VAT)
Qty:
WZ-06266-10 Nylon scraper spatula with rigid blade
Nylon scraper spatula with rigid blade
Product Type Spatulas,Material Nylon
WZ-06266-10
Usually ships in 7 days or sooner.
£53.00 GBP/EACH
£63.60 (incl VAT)
Qty:
Representative photo only NSF Approved High Heat Spatulas PP Rubber 16 1 2 L
NSF-Approved High-Heat Spatulas, PP/Rubber, 16-1/2"L
Product Type Spatulas,Material PP handle, rubber blade
WZ-06278-35
Usually ships in 7 days or sooner.
£59.00 GBP/PACKAGE
£70.80 (incl VAT)
Qty:
WZ-06283-20 Polypropylene spatula, 6 1/2
Polypropylene spatula, 6 1/2"L
Product Type Spatulas,Material Polypropylene,Sterile Yes
WZ-06283-20
Usually ships in 7 days or sooner.
£32.40 GBP/PACKAGE of 12
£38.88 (incl VAT)
Qty:
See all Spatulas, PP
WZ-06369-06 Nickel/stainless steel laboratory spatula, with 1 5/8"L flat tapered end and 1 5/8"L flat rounded end
Nickel/stainless steel laboratory spatula, with 1 5/8"L flat tapered end and 1 5/8"L flat rounded end
Product Type Spatulas,Material Nickel/Stainless Steel,Sterile Yes
WZ-06369-06
Usually ships in 7 days or sooner.
£24.19 GBP/PACKAGE of 3
£29.03 (incl VAT)
Qty:
Representative photo only Cole Parmer stainless steel micro spatula set
Cole-Parmer stainless steel micro spatula set
Product Type Spatulas,Material Stainless Steel,Brand Cole-Parmer
WZ-06369-40
Usually ships in 7 days or sooner.
£33.00 GBP/SET of 1
£39.60 (incl VAT)
Qty:
Representative photo only Stir Rod Glass 250mm 25 pk
Stir Rod, Glass, 250mm, 25/pk
Product Type Stir Rods,Material Glass
WZ-06369-23
Usually ships in 7 days or sooner.
£40.00 GBP/PACKAGE
£48.00 (incl VAT)
Qty:
See all Glass Stir Rods
WZ-06369-15 Stainless steel spatula with hardwood handle
Stainless steel spatula with hardwood handle
Product Type Spatulas,Material Stainless Steel
WZ-06369-15
Usually ships in 7 days or sooner.
£14.85 GBP/PACKAGE of 1
£17.82 (incl VAT)
Qty:
WZ-06283-30 Polypropylene spatula, 8
Polypropylene spatula, 8"L
Product Type Spatulas,Material Polypropylene,Sterile Yes
WZ-06283-30
Usually ships in 7 days or sooner.
£42.91 GBP/PACKAGE of 12
£51.49 (incl VAT)
Qty:
See all Spatulas, PP
WZ-06369-18 Nickel/stainless steel laboratory spatula, with 1 1/2"L flat squared end and 1 3/4"L flat rounded end
Nickel/stainless steel laboratory spatula, with 1 1/2"L flat squared end and 1 3/4"L flat rounded end
Product Type Spatulas,Material Nickel/Stainless Steel,Sterile Yes
WZ-06369-18
In stock.
£26.80 GBP/PACKAGE of 3
£32.16 (incl VAT)
Qty:
Representative photo only Stirring Paddle 3 Hdpe 4 cs
Stirring Paddle, 3', Hdpe 4/cs
Product Type Stir Rods,Material HDPE
WZ-06266-72
Usually ships in 7 days or sooner.
£153.00 GBP/CASE
£183.60 (incl VAT)
Qty:
WZ-06373-00 Polypropylene paddled stir rod, 9 3/4
Polypropylene paddled stir rod, 9 3/4"L
Product Type Stir Rods,Material Polypropylene
WZ-06373-00
Usually ships in 7 days or sooner.
£18.88 GBP/PACKAGE of 10
£22.66 (incl VAT)
Qty:
76 results (showing 1 - 15 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Spatulas & Stir Rods
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View Categories By: Popularity | Our Choices | All-Around Favourites  | Title | Product Category | Recommended
View popular items by: Popularity | Favourites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favourites  | Product Category | Recommended | Title