Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Pressure Regulators - Industrial
Pressure Regulators - Industrial
Pressure Regulators - Industrial
An air pressure regulator is typically used in HVAC-R systems, petrochemical processing, and slurry processing, and general manufacturing. An air pressure regulator is a vital part of applications where high-pressure fluid needs to be reduced or stabilized to a good pressure to make them safe. You can find them in various types. An air pressure regulator can be used in manufacturing using manifolds with using the regulators in line, as stand-alone units with small footprints for use in the laboratory. An air pressure regulator has a very simple operation. As the user manually changes the membrane by use of a handle, the membrane changes proportionally and allows more or less air or gas to the output connections. All regulators will have an output range for you to choose from, and various input and output pressure connections. Included brackets allow you to easily mount transducers in-line or on a panel. If you need to control any type of liquids, you will want to use a manual or electronic valve. If you need help selecting the best air pressure regulator for your applications contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Marsh Bellofram Type M2 Miniature Air Regulator, 30 psig, 1/4" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-64
In stock.
$42.00 USD/EACH
Qty:
Marsh Bellofram Type 10 Pressure Regulator, 1/4" NPT(F), 2-120 psi
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: 0.005 psi per 25 psi change psi
  • Max. supply pressure: 150 psi
WZ-68825-10
In stock.
$115.00 USD/EACH
Qty:
PVC Pressure Regulator Valve, 1/4" NPT(F)
  • Product Type: Pressure Regulating Valves
  • Material: PVC
  • Qty/ea: 1
WZ-01350-80
Available for order
or call 1-888-358-4705 to order
$517.00 USD/EACH
Qty:
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 to 30 psi
  • Product Type: Industrial Air Regulators
  • Supply pressure effect: ±0.35 psig per 25 psi psi
  • Max. supply pressure: 250 psi
WZ-68825-22
In stock.
$85.50 USD/EACH
Qty:
Miniature Regulator 0 To 10 PSIG; 1/4" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-54
In stock.
$26.75 USD/EACH
Qty:
Marsh Bellofram Type M2 Miniature Air Regulator, 120 psig, 1/4" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-68
Usually ships in 7 days.
$42.00 USD/EACH
Qty:
Marsh Bellofram Type 70 High- Flow Regulators, 1/4" NPT(F), 1 to 60 psi, 40 scfm
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: 0.025 psi
  • Max. supply pressure: 250 psi
WZ-68825-42
Available for order
or call 1-888-358-4705 to order
$95.75 USD/EACH
Qty:
Miniature Regulator 0 To 120 PSIG; 1/4" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-60
Usually ships in 7 days.
$31.25 USD/EACH
Qty:
Miniature Regulator 0 To 30 PSIG; 1/4" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-56
In stock.
$30.50 USD/EACH
Qty:
Marsh Bellofram 960-177-000, Filter Regulators, 0 to 100 psig, 40 um
  • Product Type: Industrial Air and Gas Regulators
  • Qty/ea: 1
WZ-68458-51
Usually ships in 13 days.
$64.50 USD/EACH
Qty:
Miniature Regulator 0 To 30 PSIG; 1/8" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-48
Available for order
or call 1-888-358-4705 to order
$31.25 USD/EACH
Qty:
Miniature Regulator 0 To 10 PSIG; 1/8" NPT(F)
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: <0.2 psig at 25 psi psi
  • Max. supply pressure: 145 psi
WZ-68826-46
In stock.
$31.25 USD/EACH
Qty:
Marsh Bellofram Type 70 High-Flow Regulator, 3 to 200 psi, 40 scfm
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: 0.025 psi per 25 psi psi
  • Max. supply pressure: 250 psi
WZ-68825-46
In stock.
$125.00 USD/EACH
Qty:
Marsh Bellofram Type 10 Pressure Regulator, 3/8" NPT(F), 2-120 psi
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: 0.005 psi per 25 psi change psi
  • Max. supply pressure: 150 psi
WZ-68825-16
Usually ships in 6 days.
$117.00 USD/EACH
Qty:
Marsh Bellofram Type 70 High- Flow Regulators, 3/8" NPT(F), 0 to 2 psi, 50 scfm
  • Product Type: Industrial Air and Gas Regulators
  • Supply pressure effect: 0.025 psi per 25 psi psi
  • Max. supply pressure: 250 psi
WZ-68825-48
Usually ships in 15 days.
$102.00 USD/EACH
Qty:
64 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Pressure Regulators - Industrial