Home  >  Pressure I/P Transducers
Pressure I/P Transducers
Pressure I/P Transducers
An I/P transducer changes pressure output of air and gases in direct correlation to the current or voltage input. An I/P transducer is typically used in HVAC-R systems, petrochemical processing, slurry processing, and general manufacturing. Top manufacturers include Marsh Bellofram and Brandt. The various types of transducers Cole-Parmer offers can be used in manufacturing using manifolds with using the transducers in line, as stand-alone units with small footprints for use in OEM markets and process applications, and explosion-proof units for use in petro-chemical and heavy-industrial manufacturing. Cole-Parmer also offers units with digital input and outputs. The units have a very simple operation. As current or voltage changes, a proportional pneumatic signal changes a membrane within the transducer and allows more or less air or gas to the output connections. Transducers will have an output range for you to choose from, and various input and output pressure connections. Included brackets allow you to easily mount the unit in-line or on a panel. If you need help selecting an I/P transducer or other equipment contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
WZ-68827-16 CURRENT TO PRESSURE (E/P) TRANSDUCER
Brandt STD5000, NEMA 4X I/P Transducer, 3 to 15 psi
Min operating temperature º C -40,Max operating temperature º C 66,Min operating temperature º F -40
WZ-68827-16
In stock.
$546.00 USD/EACH
Qty:
WZ-68826-00 Pressure Transducer, I to P;<br><b>3-15 PSI Output, 4-20 mA Input
Pressure Transducer, I to P;3-15 PSI Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-00
In stock.
$326.00 USD/EACH
Qty:
Representative photo only Pressure Transducer I to P 6 30 PSI Output 4 20 mA Input
Pressure Transducer, I to P;6-30 PSI Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-04
Usually ships in 7 days.
$326.00 USD/EACH
Qty:
Representative photo only Pressure Transducer I to P 3 27 PSI Output 4 20 mA Input
Pressure Transducer, I to P;3-27 PSI Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-02
Usually ships in 8 days.
$326.00 USD/EACH
Qty:
Representative photo only 2KENNG42DF31500 4 20MA 3 15 Psi Nema4X Xprf I P MB24236
2KENNG42DF31500 : 4-20MA-3-15 Psi Nema4X Xprf I/P [MB24236]
Brand Marsh Bellofram,CE Compliance Yes,Product Type I/P Transducers
WZ-68457-83
Usually ships in 15 days.
$539.00 USD/EACH
Qty:
WZ-68827-03 CURRENT TO PRESSURE (E/P) TRANSDUCER
Current to Pressure (E/P) Transducer 0 to 150 psi; 4 to 20 mA Output, 1.25 scfm
Forward flow capacity LPM 35,Min operating temperature º C 0,Max operating temperature º C 60
WZ-68827-03
Available for order.
Need it sooner?
$489.00 USD/EACH
Qty:
WZ-68827-01 CURRENT TO PRESSURE (E/P) TRANSDUCER
Current to Pressure (E/P) Transducer 0 to 30 psi; 4 to 20 mA Output, 1.25 scfm
Forward flow capacity LPM 35,Min operating temperature º C 0,Max operating temperature º C 60
WZ-68827-01
Available for order.
Need it sooner?
$489.00 USD/EACH
Qty:
WZ-68827-18 CURRENT TO PRESSURE (E/P) TRANSDUCER
Brandt STD5000, NEMA 4X I/P Transducer, 6 to 30 psi
Min operating temperature º C -40,Max operating temperature º C 66,Min operating temperature º F -40
WZ-68827-18
Available for order.
Need it sooner?
$546.00 USD/EACH
Qty:
Representative photo only Pressure Transducer I to P 4 20MA 2 60PSI I P Curr To Press
Pressure Transducer, I to P 4-20MA-2-60PSI I/P (Curr To Press)
Brand Marsh Bellofram,CE Compliance Yes,Product Type I/P Transducers
WZ-68458-26
Usually ships in 11 days.
$372.00 USD/EACH
Qty:
Representative photo only Pressure Transducer I to P 4 20MA 3 120PSI I P Curr To Press
Pressure Transducer, I to P, 4-20MA-3-120PSI I/P (Curr To Press)
Brand Marsh Bellofram,CE Compliance Yes,Product Type I/P Transducers
WZ-68458-24
Usually ships in 15 days.
$372.00 USD/EACH
Qty:
Representative photo only Pressure Trandsucer Explosion Proof I to P 6 30 psi Output 4 20 mA Input
Pressure Trandsucer, Explosion Proof, I to P;6-30 psi Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-34
Usually ships in 7 days.
$532.00 USD/EACH
Qty:
Representative photo only 2KSNNF42DF10000 4 20MA 0 100PSI Nema4X Is I P MB24235
2KSNNF42DF10000 : 4-20MA-0- 100PSI Nema4X Is I/P [MB24235]
Brand Marsh Bellofram,CE Compliance Yes,Product Type I/P Transducers
WZ-68457-82
Usually ships in 15 days.
$506.00 USD/EACH
Qty:
Representative photo only Pressure Transducer I to P 3 27 PSI Output 4 20 mA Input
Pressure Transducer, I to P;3-27 PSI Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-14
Usually ships in 7 days.
$304.00 USD/EACH
Qty:
WZ-68826-30 Pressure Trandsucer, Explosion Proof, I to P;<br><b>3-15 psi Output, 4-20 mA Input
Pressure Trandsucer, Explosion Proof, I to P;3-15 psi Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-30
Usually ships in 7 days.
$532.00 USD/EACH
Qty:
WZ-68826-06 Pressure Transducer, I to P;<br><b>3-15 PSI Output, 4-20 mA Input
Pressure Transducer, I to P;3-15 PSI Output, 4-20 mA Input
Forward flow capacity LPM 677,Min operating temperature º C -28,Max operating temperature º C 71
WZ-68826-06
Usually ships in 7 days.
$304.00 USD/EACH
Qty:
49 results (showing 1 - 15 )
Per Page 
Home  >  Pressure I/P Transducers